Skip to Content
Merck
All Photos(1)

Key Documents

568521-U

Supelco

Discovery® HS C18 (5 µm) HPLC Columns

L × I.D. 10 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L1

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

20% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

10 cm × 4.6 mm

surface area

300 m2/g

surface coverage

3.2 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Stable, low bleed for LC-MS applications
  • Scalable from analytical to preparatory
  • Highly stable to ensure excellent run-to-run and lot-to-lot reproducibility
  • Higher hydrophobicity for better resolution of difficult analytes
   

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fazli Khuda et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 944, 114-122 (2013-12-10)
A simple, specific, precise and rapid RP-HPLC-UV method was developed for simultaneous determination of lumefantrine and its metabolite desbutyl lumefantrine in human plasma. Experimental parameters were optimized and the method was validated according to standard guidelines. The method showed adequate
Lucie Zelena et al.
Journal of pharmaceutical and biomedical analysis, 137, 70-77 (2017-01-17)
Efavirenz is an antiretroviral drug used in the treatment of HIV-positive patients. A simple, fast and sensitive high-performance liquid chromatography (HPLC) method was developed in order to determine efavirenz in three types of samples provided from pharmacokinetic studies. The analysis
Abad Khan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(28), 2782-2788 (2010-09-11)
A rapid, inexpensive, sensitive and specific HPLC-ECD method for the determination of lipoic acid in human plasma was developed and validated over the linearity range of 0.001-10μg/ml using naproxen sodium as an internal standard (IS). Extraction of lipoic acid and
Jolanta Flieger et al.
Journal of separation science, 34(7), 733-739 (2011-03-02)
Highly hydrophilic compounds belonging to biogenic amines were analysed in the reversed-phase system, modified with the addition of ionic liquids: 1-ethyl-3-methyl-imidazolium hexafluorophosphate (EMIM PF(6)) and chaotropic salt NaPF(6) on Discovery HS C18 column at acidic conditions. The effect of the
Swati Jaiswal et al.
Journal of chromatographic science, 55(6), 617-624 (2017-03-24)
Tuberculosis (TB) with human immunodeficiency virus (HIV)/acquired immunodeficiency syndrome represents the most common infectious diseases worldwide. Anti-TB drugs are used concurrently with antiretroviral drug for treatment of TB-HIV co-morbidities. Due to lower risk of interaction with protease inhibitors, rifabutin is

Related Content

HPLC DiscoveryBIO HS C18 products. Available in 3µm, 5µm and 10µm particle sizes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service