Skip to Content
Merck
All Photos(3)

Documents

55269-U

Supelco

HybridSPE®-Phospholipid Ultra solid phase extraction (SPE) Cartridge

cartridge, bed wt. 30 mg, volume 1 mL, pk of 100

Synonym(s):

HybridSPE®-Phospholipid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

HybridSPE®-Phospholipid Ultra, cartridge, bed wt. 30 mg, volume 1 mL, pk of 100

material

PE frit (5-9 μm)
polypropylene hardware

Quality Level

composition

bed wt., 30 mg

packaging

pk of 100

technique(s)

solid phase extraction (SPE): suitable

volume

1 mL

matrix active group

zirconia-based phase

Looking for similar products? Visit Product Comparison Guide

General description

HybridSPE-Phospholipid technology is a simple and generic sample prep platform designed for the gross level removal of endogenous protein and phospholipid interferences from biological plasma and serum prior to LC-MS or LC-MS/MS analysis. Biological plasma or serum is first subjected to protein precipitation via the addition and mixing of acidified acetonitrile. Precipitated proteins are then removed by centrifugation and the resulting supernatant is loaded on the HybridSPE-Phospholipid 96-well plate or cartridge which acts as a chemical filter that specifically targets the removal of endogenous sample phospholipids. The phospholipid retention mechanism is based on a highly selective Lewis acid-base interaction between the proprietary zirconia ions functionally bonded to the HybridSPE-Phospholipid stationary phase and the phosphate moiety consistent with all phospholipids. The resulting eluent is ready for immediate LC-MS or LC-MS-MS analysis.

The "In-well" and "In-cartridge" precipitation methods are available for the HybridSPE-Phospholipid 96-well version and HybridSPE-Phospholipid Ultra cartridge in which biological plasma/serum is first added to either the well or cartridge, followed by acidified acetonitrile (precipitation agent). After a brief mixing/vortexing step, vacuum is applied. Because the 96-well and Ultra cartridge versions contain a series of low porosity hydrophobic filters/frits, the packed-bed filter/frit assembly acts as a depth filter facilitating the concurrent removal of both phospholipids and precipitated proteins during the extraction process. Standard HybridSPE-Phospholipid cartridges require an "off-line" precipitation method.

Features and Benefits

  • Merges the simplicity of protein precipitation and the selectivity of SPE via the targeted removal of phospholipids
  • Reduce ion-suppression through the complete removal of phospholipids and precipitated proteins
  • 2-3 step generic procedure
  • Minimal to no method development
  • Available in 96-well and 1 mL cartridge dimensions
  • Ultra cartridge allows the use of "In-cartridge" protein precipitation method
  • Filtrate is ultra clean, free of fines, and suitable for direct injection in UHPLC-type applications

Legal Information

HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of an HPLC-MS/MS method to determine clopidogrel in human plasma. Use of incurred samples to test back-conversion
Silvestro, L., et al.
Journal of Chromatography. B, Biomedical Applications, 878 (30), 3134-3142 (2010)
Solid-phase extraction of phosphorous-containing amino acid herbicides from biological specimens with a zirconia-coated silica cartridge
Watanabe, D., et al.
Journal of Chromatography. B, Biomedical Applications, 969, 69-76 (2014)
Quantitative determination of capecitabine and its six metabolites in human plasma using liquid chromatography coupled to electrospray tandem mass spectrometry
Deenen, M, et al.
Journal of Chromatography. B, Biomedical Applications, 913-914, 30-40 (2013)
C Ferreiro-Vera et al.
Journal of chromatography. A, 1240, 21-28 (2012-04-17)
Fractionation of biological fluids for proper analysis by LC-MS of low-abundance metabolites in the presence of high-abundant endogenous metabolites is of interest, particularly when the latter cause undesirable phenomena such as ionization suppression, as is the case with phospholipids (PLs).
Determination of carboplatin in human plasma using HybridSPE-precipitation along with liquid chromatography-tandam mass spectrometry
Hongliang, J,, et al.
Journal of Chromatography. B, Biomedical Applications, 879 (22), 2162-2170 (2011)

Articles

This Sigma-Aldrich article continues to detail new methodology for the analysis of Vitamin D metabolites using HybridSPE-Phospholipid technology.

This Sigma-Aldrich article continues to detail new methodology for the analysis of Vitamin D metabolites using HybridSPE-Phospholipid technology.

This Sigma-Aldrich article continues to detail new methodology for the analysis of Vitamin D metabolites using HybridSPE-Phospholipid technology.

This Sigma-Aldrich article continues to detail new methodology for the analysis of Vitamin D metabolites using HybridSPE-Phospholipid technology.

See All

Protocols

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

A simple method to enrich phospholipids from plasma samples, involving a HybridSPE-PPT 96-well plate that both retains phospholipids and removes precipitated proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service