Skip to Content
Merck
All Photos(1)

Documents

50507-U

Supelco

Ascentis® Express 90 Å C18 (5 μm) HPLC Columns

L × I.D. 2 cm × 2.1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

material

stainless steel column

Agency

suitable for USP L1

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

60 °C temp. range
600 bar max. pressure (9000 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 2.1 mm

surface area

90 m2/g

impurities

<5 ppm metals

matrix

Fused-Core particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

90 Å pore size

operating pH

2-9

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Ascentis® Express 5 μm C18 is a high-speed, high-performance liquid chromatography column based on the highly efficient Fused-Core® particle design. The Fused-Core® particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thick porous shell and the highly uniform overall particle size of 5-microns. The densely bonded, extensively endcapped dimethyloctadecyl stationary phase of Ascentis Express 5 μm C18 provides a stable, reversed-phase packing that can be used for basic, acidic, or neutral compounds.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pankaj Partani et al.
Journal of chromatographic science, 54(8), 1385-1396 (2016-05-27)
A liquid chromatography-tandem mass spectrometry method was developed and validated for the simultaneous determination of simvastatin (SV) and simvastatin acid (SVA) in human plasma. To improve assay sensitivity and achieve simultaneous analysis, SVA monitored in (-)ESI (electrospray ionization) mode within
Francesco Pio Prencipe et al.
Journal of chromatography. A, 1349, 50-59 (2014-05-27)
The study was aimed at developing a new analytical method for the metabolite fingerprinting of bioactive compounds in Humulus lupulus L. (hop), together with a simple extraction procedure. Different extraction techniques, including maceration, heat reflux extraction (HRE), ultrasound-assisted extraction (UAE)
Ahmed Abrahim et al.
Journal of pharmaceutical and biomedical analysis, 51(1), 131-137 (2009-09-18)
Fused-core silica stationary phases represent a key technological advancement in the arena of fast HPLC separations. These phases are made by fusing a 0.5 microm porous silica layer onto 1.7 microm nonporous silica cores. The reduced intra-particle flow path of
Silvia Jakabová et al.
Journal of chromatography. A, 1232, 295-301 (2012-03-07)
Hyoscyamine (atropine) and scopolamine are the predominant tropane alkaloids in the Datura genus, occurring in all plant organs. The assessment of the alkaloid content of various plant parts is essential from the viewpoint of medical use, but also as a
Heena et al.
Journal of chromatographic science, 53(5), 800-806 (2014-10-18)
This study represents a new analytical high-performance liquid chromatography-fluorescence detector method for the determination of Al(III) as Al(III) complex with 8-hydroxyquinoline-5-sulfonic acid in a tap water sample and a coke sample. A micellar liquid chromatographic method is proposed for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service