Skip to Content
Merck
All Photos(1)

Documents

48884

Supelco

ASTM® D3710 Qualitative Calibration Mix

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Agency

ASTM® D3710

CofA

current certificate can be downloaded

packaging

pkg of 6 × 1 mL

concentration

(each component in the approximate proportions (w/w) indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Butane % (w/w)
    Butylbenzene 3.2 wt. %
    Decane 3.2 % (w/w)
    2,4-Dimethylpentane 5.4 % (w/w)
    Dodecane 3.2 % (w/w)
    Heptane 9.7 % (w/w)
    Hexane 5.4 % (w/w)
    2-Methylbutane 9.7 % (w/w)
    2-Methylpentane 5.4 % (w/w)
    2-Methylpropane 1.5 % (w/w)
    Octane 5.4 % (w/w)
    Pentadecane 2.2 % (w/w)
    Pentane 7.6 % (w/w)
    Propane 1.5 % (w/w)
    Propylbenzene 4.3 % (w/w)
    Tetradecane 2.2 % (w/w)
    Toluene 10.8 % (w/w)
    Tridecane 2.2 % (w/w)
    p-Xylene 13 % (w/w)
    See All (19)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-99.9 °F - closed cup

Flash Point(C)

-73.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service