Skip to Content
Merck
All Photos(1)

Key Documents

48733

Supelco

Aroclor 1254 solution

certified reference material, 500 mg/kg in transformer oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 mL

concentration

500 mg/kg in transformer oil

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Clc1c(c(ccc1Cl)c2c(c(ccc2)Cl)Cl)Cl

InChI

1S/C12H5Cl5/c13-8-3-1-2-6(10(8)15)7-4-5-9(14)12(17)11(7)16/h1-5H

InChI key

AUGNBQPSMWGAJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing An et al.
Toxicology, 295(1-3), 8-14 (2012-03-13)
Polychlorinated biphenyls (PCBs) are still prevalent in the environment despite the fact that they have been banned in many countries for several decades. Recent epidemiologic studies have demonstrated a link between PCBs exposure and pathological alterations of bone tissues. The
L A Martin et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(3), 588-595 (2011-12-22)
Polychlorinated biphenyl (PCB) congeners differentially reduce serum thyroxine (T(4)) in rats, but little is known about their ability to affect biliary excretion of T(4). Thus, male Sprague-Dawley rats were orally administered Aroclor-1254, Aroclor-1242 (32 mg/kg per day), PCB-95, PCB-99, PCB-118
Jiali Cai et al.
Toxicology, 287(1-3), 21-28 (2011-06-15)
The testicular toxicity of polychlorinated biphenyls (PCBs) has been extensively studied. However, the detailed mechanism is still obscure. In the present study, male C57 mice were treated with different doses of Aroclor 1254 (a commercial PCB mixture) once every 3
Kyung Ho Kim et al.
Neurotoxicology, 32(6), 981-985 (2011-05-17)
Ortho-substituted polychlorinated biphenyls (PCBs) are a concern to human developmental health. Rat dams were exposed to an environmentally relevant mixture of PCBs, Aroclor 1254, or pure congener PCB 95 (6 mg/kg/day) during the perinatal period (GD 5 through PD 21).
Te-Hao Chen et al.
Ecotoxicology and environmental safety, 75(1), 207-212 (2011-09-23)
Polychlorinated biphenyls (PCBs) are a group of widespread contaminants, and accumulation of PCBs has been observed in corals in the field. However, the toxic effects of PCBs on corals have not been investigated. In this study, we tested short and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service