Skip to Content
Merck
All Photos(1)

Key Documents

48722-U

Supelco

2-Fluorobiphenyl solution

2000 μg/mL in dichloromethane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Agency

EPA 8090,8100

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Fc1c(cccc1)c2ccccc2

InChI

1S/C12H9F/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H

InChI key

KLECYOQFQXJYBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Fluorobiphenyl is generally catabolized by C. elegans, resulting in hydroxylated products. The investigation of the metabolism of 2-fluorobiphenyl by hepatic microsomes isolated from control and aroclor 1254-treated rats and pigeons has been reported.

Application

2-Fluorobiphenyl may be used as a surrogate standard for the determination of polycyclic aromatic hydrocarbons (PAHs) in soil as well as sediment samples, and pharmaceutical compounds in water samples by various chromatography techniques. It may also be used as an internal standard for the determination of organic pesticides as well as polychlorinated biphenyls (PCBs) and PAHs in sediments, water, and aquatic organisms by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reuben N Okparanma et al.
Environmental pollution (Barking, Essex : 1987), 184, 298-305 (2013-10-01)
In this study, we demonstrate that partial least-squares regression analysis with full cross-validation of spectral reflectance data estimates the amount of polycyclic aromatic hydrocarbons in petroleum-contaminated tropical rainforest soils. We applied the approach to 137 field-moist intact soil samples collected
Comparison of immunoassay and gas chromatography-mass spectrometry for measurement of polycyclic aromatic hydrocarbons in contaminated soil
Chuang JC, et al.
Analytica Chimica Acta, 486(1), 31-39 (2003)
H R Sullivan et al.
Drug metabolism and disposition: the biological fate of chemicals, 7(2), 76-80 (1979-03-01)
The absorption, metabolism, and plasma pharmacokinetics of a novel anti-inflammatory agent, 4'-ethynyl-2-fluorobiphenyl, was studied in the rat. 4'-[1-14C]Ethynyl-2-fluorobiphenyl was quantitatively absorbed from the gastrointestinal tract. Excretion of radiocarbon into urine was greater than excretion into bile. Appreciate amounts of radiocarbon
Cheng-Di Dong et al.
International journal of environmental research and public health, 9(6), 2175-2188 (2012-07-26)
Analysis of the 16 polycyclic aromatic hydrocarbons (PAHs) of the US Environmental Protection Agency priority pollutant list was carried out in sediment samples of an industrial port in the southern Kaohsiung Harbor of Taiwan which is supposed to be extensively
Compound-Specific Gas Chromatographic/Mass Spectrometric Analysis of Alkylated and Parent Poly cyclic Aromatic Hydrocarbons in Waters, Sediments, and Aquatic Organisms
Means JC
Journal of AOAC (Association of Official Analytical Chemists) International, 81(3), 657-672 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service