Skip to Content
Merck
All Photos(1)

Key Documents

48624

Supelco

Bromomethane solution

certified reference material, 200 μg/mL in methanol

Synonym(s):

Methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Br
CAS Number:
Molecular Weight:
94.94
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CBr

InChI

1S/CH3Br/c1-2/h1H3

InChI key

GZUXJHMPEANEGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromomethane is a broad spectrum fumigant, which can find applications in post-harvest fumigation and pest control of several foods like spices, cereals, nuts, dried fruits, fresh fruits, vegetables, turf and also nursery crops.
Bromomethane is frequently used as fumigant. It is a liquid at room temperature which readily vaporizes into a colourless gas. It is slightly soluble in water and is freely soluble in organic solvents.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Solution of bromomethane may be used in preparation of 2-methoxyisoindoline-1,3- dione.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Handbook of Chemicals and Safety (2016)
The challenge of research and extension to define and implement alternatives to methyl bromide
Noling.W.J and Becker.OJ
Journal of Nematology, 26, 573-573 (1994)
Fortune Ezemobi et al.
Acta crystallographica. Section E, Structure reports online, 70(Pt 12), 456-461 (2015-01-02)
The title compounds, C9H7NO3, (1), C10H7NO5, (2), and C14H9NO5, (3), are three potentially anti-convulsant compounds. Compounds (1) and (2) are isoindoline derivatives and (3) is an iso-quinoline derivative. Compounds (2) and (3) crystallize with two independent mol-ecules (A and B)
Khristy J Thompson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 120(1), 163-172 (2010-12-24)
Manganese (Mn) is an essential nutrient with potential neurotoxic effects. Mn deposited in the nose is apparently transported to the brain through anterograde axonal transport, bypassing the blood-brain barrier. However, the role of the olfactory epithelial cells in Mn transport
Kathryn Hughes Barry et al.
Cancer causes & control : CCC, 23(6), 807-818 (2012-04-25)
Methyl bromide is a genotoxic soil fumigant with high acute toxicity, but unknown human carcinogenicity. Although many countries have reduced methyl bromide use because of its ozone depleting properties, some uses remain in the United States and other countries, warranting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service