Skip to Content
Merck
All Photos(1)

Key Documents

47330-U

Supelco

PCB Congener Mix 1

10 μg/mL each component in isooctane, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 10 mL

concentration

10 μg/mL each component in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Application

PCB Congener Mix 1 is used as an analytical standard for the post degradation analysis of polychlorinated biphenyls (PCBs) in dry fermented sausages[1] and in meat emulsions of a frankfurter type[2] using gas chromatography-mass spectrometry technique[1] and gas chromatography (GC)-micro electron capture detector (μECD).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    2,6-Dichlorobiphenyl
    2,2′,3,4,4′,5,5′-Heptachlorobiphenyl
    2,2′,3,4,4′,5′-Hexachlorobiphenyl
    2,2′,4,4′,5,5′-Hexachlorobiphenyl
    2,2′,5,5′-Tetrachlorobiphenyl
    2,4,4′-Trichlorobiphenyl

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of PCBs in a frankfurter-type meat emulsion: Effects of a meat starter, its proteins extract and thermal treatments
Lusnic M, et al.
Food And Chemical Toxicology, 50(8), 2643-2647 (2012)
Degradation of PCBs in dry fermented sausages during drying/ripening
Polak LM, et al.
Food Chemistry, 213, 446-250 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service