Skip to Content
Merck
All Photos(1)

Key Documents

47133

Supelco

β-Sitosterol

certified reference material, 100 μg/mL in chloroform

Synonym(s):

α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Beilstein:
1916165
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

100 μg/mL in chloroform

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

136-140 °C (lit.)

application(s)

food and beverages

format

single component solution

functional group

hydroxyl

storage temp.

-10 to -25°C

SMILES string

O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@H](C(C)C)CC

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuntakorn Thongtang et al.
Atherosclerosis, 225(2), 388-396 (2012-10-09)
Statins inhibit cholesterol synthesis but can upregulate cholesterol absorption, with higher doses producing larger effects. Ezetimibe inhibits cholesterol absorption but also upregulates synthesis. We tested whether ezetimibe added to on-going statin therapy would be most effective in lowering LDL-cholesterol (LDL-C)
Daniele Silvestro et al.
PloS one, 8(2), e56429-e56429 (2013-02-15)
Sterols are crucial lipid components that regulate membrane permeability and fluidity and are the precursors of bioactive steroids. The plant sterols exist as three major forms, free sterols, steryl glycosides and steryl esters. The storage of steryl esters in lipid
Günther Silbernagel et al.
Current opinion in lipidology, 24(1), 12-17 (2012-11-21)
Plant sterols as ingredients to functional foods are recommended for lowering LDL cholesterol. However, there is an ongoing discussion whether the use of plant sterols is safe. Genetic analyses showed that common variants of the ATP binding cassette transporter G8
Ashutosh Singh
Food chemistry, 137(1-4), 62-67 (2012-12-04)
The antioxidative effect of sitosterol at 1, 2 and 5% levels, in triolein, refined canola, high oleic sunflower and flaxseed oils, continuously heated for a period of up to 72 h at frying temperature of 180 °C, was studied. High
M L Jones et al.
European journal of clinical nutrition, 66(11), 1234-1241 (2012-09-20)
The percentage of hypercholesterolemic individuals not reaching their LDL-cholesterol (LDL-C) goal remains high and additional therapeutic strategies should be evaluated. The objective of this study was to evaluate the cholesterol-lowering efficacy and mechanism of action of bile salt hydrolase-active Lactobacillus

Protocols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service