Skip to Content
Merck
All Photos(1)

Documents

47112-U

Supelco

Corn oil

analytical standard

Synonym(s):

Maize oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

This reference material is offered as a qualitative standard. Use the chromatographic fingerprint of our reference material to check for the presence of corn oil in your sample. A Certificate of Composition which includes a chromatogram is shipped with each product. Characterized corn oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen. A Certificate of Composition, which includes the chromatographic fingerprint analysis, is provided with the oil sample.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Corn oil is used as a characterized reference standard to investigate its fatty acid composition using gas chromatography technique after subjecting it to transesterification reaction.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

489.2 °F - open cup

Flash Point(C)

254 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aimin Shi et al.
Bioresource technology, 128, 100-106 (2012-12-01)
Six Korea high oil (KHO) corn varieties varying in germ and endosperm size and oil content (4-21%, wet basis) were subjected to three sequential combinations of milling (M), germ separation (S), fermentation (F), and in situ transesterification (T) to produce
Yingyi Mao et al.
Journal of food science, 77(5), E144-E152 (2012-11-21)
This study describes how the rheological properties of colloidal dispersions formed by heteroaggregation of oppositely charged protein-coated lipid droplets depend on total particle concentration. Mixed-particle emulsions were formed by mixing single-particle emulsions containing either β-lactoglobulin-coated lipid droplets (ζ≈-42 mV, d(43)≈
Hui Li et al.
Nature genetics, 45(1), 43-50 (2012-12-18)
Maize kernel oil is a valuable source of nutrition. Here we extensively examine the genetic architecture of maize oil biosynthesis in a genome-wide association study using 1.03 million SNPs characterized in 368 maize inbred lines, including 'high-oil' lines. We identified
Hua-Min Liu et al.
Bioresource technology, 128, 58-64 (2012-12-01)
Hydrothermal liquefaction of cornstalk at 180-300 °C at ratios of water to cornstalk of 6-14 was conducted, and the reaction products were lumped into gas, water-soluble organics (ethanol-insoluble and ethanol-soluble organics), heavy oil, volatile organic compounds, and acid-soluble and acid-insoluble
B G Kim et al.
Journal of animal science, 91(2), 755-763 (2012-11-14)
An experiment was conducted to determine the true ileal digestibility (TID) and the true total tract digestibility (TTTD) of acid-hydrolyzed ether extract (AEE) in extracted corn oil, high-oil corn, distillers dried grains with solubles (DDGS), corn germ, and high protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service