Skip to Content
Merck
All Photos(1)

Documents

46940-U

Supelco

D-Malic acid

analytical standard

Synonym(s):

D-(+)-Malic acid, (R)-(+)-2-Hydroxysuccinic acid, D-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723540
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E296
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

98-102 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

O[C@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m1/s1

InChI key

BJEPYKJPYRNKOW-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siva S V P Sakamuri et al.
GeroScience, 40(3), 347-356 (2018-06-04)
Mitochondria play a critical role in the cardiomyocyte physiology by generating majority of the ATP required for the contraction/relaxation through oxidative phosphorylation (OXPHOS). Aging is a major risk factor for cardiovascular diseases (CVD) and mitochondrial dysfunction has been proposed as
Duoduo Chen et al.
Molecular microbiology, 87(6), 1237-1244 (2013-01-26)
Nicotine is an environmental toxicant in tobacco waste, imposing a serious hazard for human health. Some bacteria including Pseudomonas spp. strains are able to metabolize nicotine to non-toxic compounds. The pyrrolidine pathway of nicotine degradation in Pseudomonas putida S16 has
Narendra Kumar Sharma et al.
Journal of proteomics, 79, 277-298 (2013-01-15)
Hypoxia is one of the major stressors at high altitude. Exposure to hypobaric hypoxia induces several adverse consequences to the structural and functional integrity of brain. In an attempt to understand the proteome modulation, we used 2-DE coupled with MALDI-TOF/TOF
Cuiyue Liang et al.
Plant physiology, 161(3), 1347-1361 (2013-01-24)
Low pH, aluminum (Al) toxicity, and low phosphorus (P) often coexist and are heterogeneously distributed in acid soils. To date, the underlying mechanisms of crop adaptation to these multiple factors on acid soils remain poorly understood. In this study, we
E González-Fandos et al.
Poultry science, 92(7), 1936-1941 (2013-06-19)
This work evaluated the effect of malic acid washing on the growth of Listeria monocytogenes on poultry legs stored at 4°C for 8 d. Fresh inoculated chicken legs were dipped into a 1 or 2% malic acid solution (vol/vol) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service