Skip to Content
Merck
All Photos(1)

Key Documents

40025

Supelco

1,4-Dichlorobenzene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)Cl

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

OCJBOOLMMGQPQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanako Watanabe et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 25(11), 1301-1305 (2009-11-13)
A very simple and sensitive method for the simultaneous analysis of naphthalene and p-dichlorobenzene in human whole blood and urine by headspace capillary gas chromatography-mass spectrometry (GC-MS) is presented. The advantages of the method were that as much as 1
1,4-Dichlorobenzene.
Report on carcinogens : carcinogen profiles, 12, 139-141 (2011-08-19)
Gayathri D Warnasuriya et al.
Archives of toxicology, 84(3), 233-243 (2010-01-14)
Short-chain chlorinated paraffins (SCCPs) cause kidney tumours in male rats, but not in female rats or mice of either sex. Male rat-specific tumours also occur in rats dosed with a range of compounds including 1,4- dichlorobenzene (DCB) and d-limonene (DL).
Case files of the New York City poison control center: paradichlorobenzene-induced leukoencephalopathy.
Stephanie H Hernandez et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 6(2), 217-229 (2010-04-08)
Banu Sizirici et al.
Waste management (New York, N.Y.), 30(1), 82-91 (2009-10-06)
The purpose of this study was to evaluate suitability of using the time series analysis for selected leachate quantity and quality parameters to forecast the duration of post closure period of a closed landfill. Selected leachate quality parameters (i.e., sodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service