23906
OMI® Purifier Tube
OMI-2 Purifier Tube
Synonym(s):
OMI®-2 INDICATOR TUBE
About This Item
Recommended Products
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
General description
- A re-useable tube holder that is installed into the gas delivery system. The use of polycarbonate provides see-through capability along with safety.
- A purifier tube containing an indicating resin (changes color from black to brown when exposed to as little as 1 ppm of moisture or oxygen). The use of glass provides see-through capability and prevents diffusion of room contaminants into the gas stream.
Spent purifier tubes are easily replaced. Simply unscrew the end assembly from the tube holder and replace it with a new purifier tube. The design prevents room air from entering the new tube during installation (protective foil covers on each end are only pierced as the end assembly is screwed back onto the tube holder to complete installation).
Note: This item requires a re-useable tube holder (purchased separately).
Footnote
Legal Information
required but not provided
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - Water-react 1
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.
GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.
GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.
GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.
Related Content
Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.
Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service