Skip to Content
Merck
All Photos(4)

Documents

23094-U

Supelco

1/4 in. Brass Swagelok® and PTFE Ferrule End-Fitting Assembly, Precleaned for Thermal Desorption Tube

greener alternative

for use with 1/4 in. (6.35 mm) Thermal Desorption Tube, pk of 20 ea

Synonym(s):

Pre-cleaned Brass Swagelok and PTFE Ferrule Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

brass (precleaned and assembled)

product line

Swagelok®

packaging

pk of 20 ea

manufacturer/tradename

Swagelok® B-400-C

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

fitting

1/4 in. Swagelok

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with 1/4 in. (6.35 mm) Thermal Desorption Tube

greener alternative category

General description

The assembly includes a 1/4" brass Swagelok® and 1/4 in. PTFE ferrule, pre-cleaned and ready for use. This assembly is ideal for those who purchase unconditioned glass and stainless steel thermal desorption tubes and condition in their own lab.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service