Skip to Content
Merck

Skip To

22012

SWAGELOK® Front Ferrule

SWAGELOK® 203-1, brass, 1/8 in. SWAGELOK

Synonym(s):

Swagelok® 203-1,BRASS FRT FERR 1/8" PK20

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41115717
NACRES:
SB.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

brass

packaging

pkg of 20 ea

manufacturer/tradename

SWAGELOK® 203-1

fitting

1/8 in. SWAGELOK

General description

Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing.

Application

Brass front ferrule maybe used for sealing the tube ends to increase the lifetime and performance of the permeation tubes as standard gas sources.

Legal Information

SWAGELOK is a registered trademark of Swagelok Company

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
220382202722039
fitting

1/8 in. SWAGELOK

fitting

1/8 in. SWAGELOK

fitting

1/4 in. SWAGELOK

fitting

1/8 in. SWAGELOK

packaging

pkg of 20 ea

packaging

pkg of 5 ea

packaging

pkg of 5 ea

packaging

pkg of 5 ea

manufacturer/tradename

SWAGELOK® 203-1

manufacturer/tradename

SWAGELOK® 203-1

manufacturer/tradename

SWAGELOK® 403-1

manufacturer/tradename

SWAGELOK® 204-1

material

brass

material

stainless steel

material

stainless steel

material

stainless steel


Still not finding the right product?

Explore all of our products under Swagelok® Front Ferrule




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters