Skip to Content
Merck
All Photos(1)

Documents

21342-U

Supelco

Washed Silica Gel

pkg of 250 g bulk packing

Synonym(s):

Cleaned Silica Gel for EPA 1631

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

Agency

suitable for EPA 1613

Quality Level

form

solid

packaging

pkg of 250 g bulk packing

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

Application


  • Electroanalytical sensing of trichloroacetic acid based on activated self-supported porous silver wire prepared by voltammetric etching with trichloroacetic acid as etchant.: This study presents an innovative method for detecting trichloroacetic acid (TCA) using a self-supported porous silver wire. The electrochemical sensor demonstrates high sensitivity and selectivity for TCA, making it a valuable tool in environmental monitoring and water quality assessment (Chu et al., 2022).



Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Otmar Geiss et al.
Journal of chromatography. A, 1566, 13-22 (2018-06-28)
Polycyclic Aromatic Hydrocarbons (PAHs) have been detected in rubber and plastic components of a number of consumer products such as toys, tools for domestic use, sports equipment, and footwear, with carbon black and extender oils having been identified as principal

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service