Skip to Content
Merck
All Photos(1)

Key Documents

10286

HayeSep® Porous Polymer Adsorbent

matrix HayeSep B, 80-100 mesh, bottle of 75 cc

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23201100
eCl@ss:
32119290

form

solid

Quality Level

packaging

bottle of 75 cc

manufacturer/tradename

Hayes Separation Inc

parameter

190 °C temp. limit

technique(s)

gas chromatography (GC): suitable

surface area

~608 m2/g

matrix

HayeSep B

particle size

80-100 mesh

density

~0.33 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water.They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. HayeSep® has a high surface area owing to its micropores, thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). Furthermore they are relatively inert and exhibit hydrophobicity. HayeSep B is a copolymer of divinylbenzene and polyethylene.
HayeSep porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Application

HayeSep B can maybe used in combination with HayeSep A for separation of a gaseous mixture comprising of hydrogen and hydrogen sulphide.

Legal Information

HayeSep is a registered trademark of Hayes Separation Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas Chromatography
Poole C. et al.
Science, 127-130 (2012)
Specific features of the gas-chromatographic determination of the reaction products in the catalytic decomposition of hydrogen sulfide.
Zheivot V, et al.
Journal of Analytical Chemistry, 61 (4), 334-337 (2006)
Trace Analysis of Specialty and Electronic Gases.
Geiger W.M and Raynor M.W. et al.
Science, 256-257 (2013)
Gas Chromatography in Air Pollution Analysis.
Berezkin V.G and Drugov Y.S. et al.
J. Chromatogr. Library, 49, 50-51 (1991)
Modern Practice of Gas Chromatography.
Grob RL and Barry EF. et al.
Science, 79-83 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service