Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1454

Sigma-Aldrich

Anti-TIGIT Antibody, clone 1D20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

T-cell immunoreceptor with Ig and ITIM domains, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain-containing protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D20, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 26.32 kDa
observed mol wt 40 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TIGIT(201633)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D20 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human T-cell immunoreceptor with Ig and ITIM domains (TIGIT). It targets an epitope within 16 amino acids from the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal, extracellular domain of human T-cell immunoreceptor with Ig and ITIM domains (TIGIT).

Application

Anti-TIGIT, clone 1D20 ZooMAb, Cat. No. ZRB1454, is a recombinant Rabbit monoclonal antibody that specifically targets TIGIT and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TIGIT in Rat Spleen, Mouse Thymus, and Human Spleen tissue lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TIGIT in Jurkat and K562 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TIGIT in human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound TIGIT peptide with a KD of value of 3.6 x 10-9 in an affinity binding assay.

Flow Cytometry Analysis: 1 µg of a representative lot of this antibody detected TIGIT in one million Raw264.7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

T-cell immunoreceptor with Ig and ITIM domains (UniProt: Q495A1; also known as V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain-containing protein 3, TIGIT) is encoded by the TIGIT (also known as VSIG9, VSTM3) gene (Gene ID: 201633) in human. TIGIT is a single-pass type I membrane protein that binds with high affinity to the poliovirus receptor (PVR) forming a heterotetrameric assembly of two TIGIT and two PVR molecules. This causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T-cell activation by promoting the generation of mature immunoregulatory dendritic cells. It is expressed at low levels on peripheral memory and regulatory CD4+ T-cells and NK cells and is up-regulated following activation of these cells. It is also expressed on many cell types important in cancer, and its mechanism of action may depend on the cell type studied. TIGIT also serves as a functional ligand and induces dendritic cells to acquire a tolerogenic phenotype via ligation of PVR. TIGIT -/- mice, when challenged, develop severe experimental autoimmune encephalomyelitis compared to wild-type mice. TIGIT is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 22-141), a transmembrane domain (aa 142-162), and a cytoplasmic domain (aa 163-244). The cytoplasmic domain contains an immunoreceptor tyrosine-based inhibitor motif (ITIM; aa 229-234), which is involved in the modulation of cellular responses. The IgV domain (aa 22-124) of TIGIT possesses distinctive (V/I)(S/T)Q, AX6G, and T(F/Y)PX1G sub-motifs that mediate a ′lock and key′ trans-interaction with cis-homodimers of PVR. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Manieri, NA., et al. (2017). Trends Immunol. 38(1); 20-28).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service