Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1284

Sigma-Aldrich

Anti-c-Kit/CD117 Antibody, clone 1L19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 109.87 kDa
observed mol wt ~160 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L19 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects c-Kit/CD117. It targets an epitope within 23 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal, extracellular domain of human c-Kit/CD117.

Application

Quality Control Testing

Evaluated by Western Blotting in TF-1 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected c-Kit/CD117 in TF-1 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected c-Kit/CD117 in NIH3T3 cell lysate.

Flow Cytometry Analysis: 1 μg from a representative lot detected c-Kit/CD117 in one million K562 cells.

Affinity Binding Assay: A representative lot of this antibody bound CD117/c-Kit with a KD of 1.8 x 10-7in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1000 dilution from a representative lot detected c-Kit/CD117 in TF-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-c-Kit/CD117, clone 1L19 ZooMAb®, Cat. No. ZRB1284, is a recombinant Rabbit monoclonal antibody that specifically targets v-Kit/CD117 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, and Western Blotting.

Target description

Mast/stem cell growth factor receptor Kit (UniProt: P10721; also known as EC: 2.7.10.1, SCFR, Piebald trait protein, PBT, Proto-oncogene c-Kit, Tyrosine-protein kinase Kit, p145 c-kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog) is encoded by the KIT (also known as SCFR) gene (Gene ID: 3815) in human. c-Kit is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 26-524), a transmembrane domain (aa 525-545), and a cytoplasmic domain (aa 546-976). c-Kit has tyrosine protein kinase activity and serves as a cell-surface receptor for the stem cell factor (SCF)/KITLG. It plays a significant role in the regulation of cell survival, cell proliferation, stem cell maintenance, gametogenesis and other functions. In response to SCF binding, c-Kit can activate several signaling pathways. In the absence of bound stem cell factor (SCF), it exists as a monomer, however, it homodimerizes in the presence of bound SCF and forms a heterotetramer with two SCF molecules. c-Kit can be autophosphorylated on tyrosine residues and SCF binding enhances its autophosphorylation. Its kinase activity is reported to be down-regulated by phosphorylation on serine residues by protein kinase C family members. Phosphorylation at tyrosine 568 is shown to be essential for its interaction with PTPN11/SHP-2, CRK (isoform Crk-II) and members of the Src tyrosine-protein kinase family and phosphorylation at tyrosine 570 is required for interaction with PTPN6/SHP-1. c-Kit can be ubiquitinated by SOCS6 and it is rapidly ubiquitinated after autophosphorylation induced by SCF binding, leading to its internalization and degradation. Lack of c-Kit expression has been reported in multiple cancer types and c-Kit expressing cells display a regression of typical morphological features of malignancy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service