Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1113

Sigma-Aldrich

Anti-LILRB4 Antibody, clone 1H12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H12, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~49.36 kDa
observed mol wt ~50 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects human Leukocyte immunoglobulin-like receptor subfamily B member 4 (LILRB4). It targets an epitope within 19 amino acids from the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the extracellular domain of human Leukocyte immunoglobulin-like receptor subfamily B member 4 (LILRB4).

Application

Quality Control Testing

Evaluated by Western Blotting in MDA-MB-435 lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LILRB4 in MDA-MB-435 cell lysate.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LILRB4 in NK-92 cells.

Affinity Binding Assay: A representative lot of this antibody bound LILRB4 peptide with a KD of 2.5 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 1 μg from a representative lot detected LILRB4 in NK-92 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LILRB4, clone 1H12 ZooMAb®, Cat. No. ZRB1113, is a rabbit recombinant monoclonal antibody that specifically targets LILRB4 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, and Western Blotting.

Target description

Leukocyte immunoglobulin-like receptor subfamily B member 4 (UniProt: Q8NHJ6; also known as CD85 antigen-like family member K, Immunoglobulin-like transcript 3, ILT-3, Leukocyte immunoglobulin-like receptor 5, LIR-5, Monocyte inhibitory receptor HM18, CD85k, LILRB4) is encoded by the LILRB4 (Gene ID: ILT3, LIR5) gene (Gene ID: 11006) in human. LILRB4 is a single-pass type I membrane protein that serves as a receptor for class I MHC antigens and recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles. It is involved in the downregulation of the immune response and the development of tolerance, such as towards transplants. It is reported to inhibit receptor-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions. LILRB4 expression has been reported in monocytes, macrophages, dendritic cells, lung, natural killer cells and B-cells and is induced upon contact with CD8+ CD28- alloantigen-specific T suppressor (Ts) cells. LILRB4 is also a tumor-associated antigen that is highly expressed on monocytic AML cells where its expression is more specific and can be used to differentiate monocytic AML (M5) from AML M1-M3. It is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 22-259), a transmembrane domain (aa 260-280), and a cytoplasmic domain (aa 181-448). It contains three copies of immunoreceptor tyrosine-based inhibitor motif (ITIM) that are involved in modulation of cellular responses and the phosphorylated ITIM motif can bind the SH2 domain of several SH2-containing phosphatases. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: John, S. et al. (2018). Mol. Therap. 26(10); 2487-2495).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service