Skip to Content
Merck
All Photos(14)

Key Documents

ZMS1122

Sigma-Aldrich

Anti-MRP-8/14 Antibody, clone AHN-17 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein S100-A8/9;MRP-8/MRP-14;Leukocyte L1 complex light chain/ Leukocyte L1 complex heavy chain;p8/p14;S100 calcium-binding protein A8/A9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ELISA
ICC
IHC
affinity binding assay
clone:
AHN-17, recombinant monoclonal
species reactivity:
human
citations:
technique(s):
ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

AHN-17, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 13.24 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone AHN-17 is a ZooMAb® Mouse recombinant monoclonal antibody that specifically detects MRP-8/MRP-14 (Protein S100-A8/9).

Immunogen

Human Neutrophils.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected MRP-8/14 in human tonsil tissue sections.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound recombinant human MRP-8/14 protein with a KD of 8.7 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): Serial dilutions from a representative lot detected recombinant human MRP-8/14.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MRP-8/14 in HL-60 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected MRP-8/14 in human tonsil tissue sections.

Target description

Protein S100-A8/9 (UniProt: P05109/ P06702; also known as MRP-8/MRP-14, Leukocyte L1 complex light chain/ Leukocyte L1 complex heavy chain, p8/p14, S100 calcium-binding protein A8/A9) are encoded by the S100A8/S100A9 (also known as MRP8/MRP14) genes (Gene ID: 6279/6280) in human. MRP-8 and MRP-14 are calcium- and zinc-binding proteins that play a prominent role in the regulation of inflammatory processes and immune response. They are known to induce neutrophil chemotaxis and adhesion. They are predominantly found as calprotectin and perform a variety of intra- and extracellular functions. Their intracellular functions include facilitating leukocyte arachidonic acid trafficking and metabolism, modulation of the tubulin-dependent cytoskeleton during migration of phagocytes, and activation of the neutrophilic NADPH-oxidase. They are known to activate NADPH-oxidase by facilitating the enzyme complex assembly at the cell membrane. MRP-8 contributes to the enzyme assembly by directly binding to NCF2/P67PHOX. Its extracellular functions include proinflammatory, antimicrobial, oxidant-scavenging and induction of apoptosis. MRP-14 is reported to induce neutrophil chemotaxis and adhesion and can increase the bactericidal activity of neutrophils by promoting phagocytosis via activation of SYK, PI3K/AKT, and ERK1/2 pathways and can induce degranulation of neutrophils by a MAPK-dependent mechanism. MRP-8 and MRP-14 also acts as danger associated molecular pattern (DAMP) molecules and stimulate innate immune cells via binding to pattern recognition receptors such as Toll-like receptor 4 (TLR4) and receptor for advanced glycation end (AGER) products. They also play an impotent role as oxidant scavengers and have a protective role in preventing exaggerated tissue damage by scavenging oxidants. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Atallah, M., et al. (2012). PloS One. 7(2); e29333; Nakatani, Y., et al. (2005). Mediators Inflamm. 2005(5); 280-292; Ryckman, C., et al. (2003). J. Immunol. 170(6); 3233-3242).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service