Skip to Content
Merck
All Photos(1)

Key Documents

Z740250

HotSeal sealing film for PCR applications

sterile

Synonym(s):

PCR plate sealing film, multiplate sealing film, plate seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22
Pricing and availability is not currently available.

material

polyester film

sterility

sterile

packaging

pkg of 100 ea (sheets)

manufacturer/tradename

Diversified Biotech HOTS-1000

technique(s)

PCR: suitable

General description

  • Suitable for PCR applications.
  • Free of RNase and DNase.
  • Provided with a thick polyester film of strength 2.3 mil
  • Two scored end tabs enable easy positioning.
  • Ideal temperature range: -40°C to 125°C
  • Eliminates well-to-well contamination, evaporation, and spillage.
  • Neither gas permeable nor UV transparent.

Legal Information

HaackerCalc is a trademark of ChemSW, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service