Z740229
TLC Aluminum Sheets
L × W 5 cm × 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indicator
Synonym(s):
Aluminum Backed TLC Plates
About This Item
Recommended Products
material
aluminum support
silica gel 60 matrix
Quality Level
feature
binder Polymeric
fluorescent indicator
packaging
pkg of 100 ea
manufacturer/tradename
EMD Millipore® 1.05534.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
5 cm × 20 cm
layer thickness
200 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service