Skip to Content
Merck
All Photos(1)

Key Documents

Z709220

ART® self-sealing barrier pipette tips

ART XLG, volume range 5-200 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
200 μL
volume range:
5-200 μL
sterility:
sterile
feature:
barrier
packaging:
pack of 3072 ea (4 packs of 768)
pkg of 768 tips (racked)
manufacturer/tradename:
Molecular BioProducts 2160G

material

colorless tip

sterility

sterile

feature

barrier

packaging

pack of 3072 ea (4 packs of 768)
pkg of 768 tips (racked)

manufacturer/tradename

Molecular BioProducts 2160G

technique(s)

PCR: suitable

maximum volume

200 μL

volume range

5-200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Extended length and large orifice tip for handling fragile cells, viscous samples and genomic DNA

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit Proline-Variable 50-200 μl
Capp/Aero 200 μl
Eppendorf Research/Reference 20-200 μl
Eppendorf 4810 10-100 μl
Eppendorf Research Pro 5-100 μl
Eppendorf Response 4850 5-100 μl
Finnpipette 4027/4500 5-40 μl
Mettler-Toledo VoluMate 20-200 μl
Nichiryo BenchMate 20-200 μl
Pipetman P-200 20-200 μl
Rainin EDP 2 20-250 μl
Rainin EDP Plus 20-250 μl
Rainin Pipet Plus 20-200 μl
Rainin Pipetman 20-200 μl
Socorex/Wheaton 822 Series 10-100 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service