Before autoclaving catalog number Z380237, completely remove closure and set on top of the container at an angle, without engaging the threads.
Z380237
Jars Nalgene® style 2118
capacity 125 mL
Synonym(s):
laboratory jars, nalgene jars
About This Item
Recommended Products
material
polypropylene bottle
polypropylene cap
round translucent bottle
Joint
Joints threaded neck (70)
Agency
suitable for EPA OTM-45
description
Autoclavable with closure completely unscrewed
feature
with cap
packaging
pkg of 12
manufacturer/tradename
Nalgene® 2118-0004
capacity
125 mL
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
-
How do I autoclave Product Z380237, Jars Nalgene® style 2118?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
Can you autoclave Product Z380237, Jars Nalgene® style 2118, more than one time?
1 answer-
Yes. Catalog number Z380237 can be autoclaved repeatedly, with excellent chemical resistance.
Helpful?
-