Key Documents
Z374938
Micro mats for PCR plates
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
technique(s)
PCR: suitable
Related Categories
General description
  • Fits most popular 96-well PCR plates
  • Minimized evaporation in PCR and storage
  • Alpha numerics printed on each mat, for easy well identification
  • Pierceable with syringe
  • Not compatible with DMSO and other harsh solvents
  • Tabs for easy application/removal of mat
  • Reusable for up to 5 times
  • When used with a screw- or clip-down thermal cycler lid, provides 100% sealing
  • Fully autoclavable, reversible
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.
See All