Skip to Content
Merck
All Photos(1)

Key Documents

Z2876

Sigma-Aldrich

Zinc sulfate solution

0.3 N

Synonym(s):

Zinc Sulfate hydrate, ZnSO4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4
CAS Number:
Molecular Weight:
161.45
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
aqueous solution

grade

for analytical purposes

form

aqueous solution

concentration

0.3 N

SMILES string

[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Zn/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

NWONKYPBYAMBJT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in conjunction with barium hydroxide to deproteinize via barium sulfate precipitation whole blood, plasma, or serum samples which are colored or turbid. The resulting supernatant or filtrate is then available for subsequent analysis of glucose by Somogi-Nelson method.

related product

Product No.
Description
Pricing

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luiz Osório Leiria et al.
Cell metabolism, 30(4), 768-783 (2019-07-30)
Distinct oxygenases and their oxylipin products have been shown to participate in thermogenesis by mediating physiological adaptations required to sustain body temperature. Since the role of the lipoxygenase (LOX) family in cold adaptation remains elusive, we aimed to investigate whether
Zinc sulfate treatment of vocal process granuloma.
Taner Yilmaz
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(3), 1175-1175 (2012-10-12)
Acute diarrhea in adults and children: a global perspective.
Michael Farthing et al.
Journal of clinical gastroenterology, 47(1), 12-20 (2012-12-12)
Dervis Özcelik et al.
Biological trace element research, 150(1-3), 342-349 (2012-10-12)
Zinc is an element that under physiological conditions preferentially binds to and is a potent inducer of metallothionein under physiological conditions. The present study was conducted to explore whether zinc supplementation morphologically and biochemically protects against diabetic nephropathy through modulation
O I Smian et al.
Likars'ka sprava, (3-4)(3-4), 67-71 (2013-01-30)
The article analyzed 60 patients with community-aquired pneumonia from ages 6 to 18 years. The dependences of trace levels of zinc and copper in the serum of these children on the activity of the inflammatory process. Analysis of survey results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service