Key Documents
WHAWB100076
Unicore Punch Kit
I.D. 2.00 mm, pkg of 25 ea
Synonym(s):
UniCore Punches 2.00 mm (25), Whatman FTA, Z708844, harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
Pricing and availability is not currently available.
packaging
pkg of 25 ea
manufacturer/tradename
Qiagen WB100076
I.D.
2.00 mm
General description
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Manual UniCore punch system for sample disc removal from FTA cards.
Features and Benefits
  • Stainless steel cutting tip (ranging from 2.0 mm) enables manual coring, retrieval, and controlled ejection of the FTA punch.
  • Designed to minimize contamination and carryover between samples when a cleaning procedure is followed.
Other Notes
Field of Use : For internal research use only
Legal Information
FTA is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.
This document shows intructions for EasiCollect™ device from Cytiva.