WHA77002801
Whatman® UNIFILTER® 96 well filter plates
clear polystyrene, GF/C membrane, non-sterile, biomolecule, 25 ea
Synonym(s):
Z382310, micro plate, multiwell plate, well plates
About This Item
Product Name
Whatman® UNIFILTER® plates, working volume 800 μL, with Whatman GF/C membrane
material
clear polystyrene
conical bottom clear wells
polystyrene filter
sterility
non-sterile
packaging
pkg of 25 ea
manufacturer/tradename
Cytiva
Whatman 7700-2801
Whatman Article No. 28420805 (US reference)
maximum volume
800 μL
size
96 wells
working volume
800 μL
binding type
non-treated surface
Related Categories
Features and Benefits
- No cross talk. Patented integral filter design prevents well-to-well cross contamination.
- Economical to use. Wide range of well volume options ensures efficient use of materials.
- Better control. Choice of filter media allows control of the flow rates and retention characteristics.
- Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)