Skip to Content
Merck
All Photos(2)

Documents

WH0375611M4

Sigma-Aldrich

Monoclonal Anti-SLC26A5 antibody produced in mouse

clone 1F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DFNB61, Anti-MGC118886, Anti-MGC118887, Anti-MGC118888, Anti-MGC118889, Anti-PRES, Anti-solute carrier family 26, member 5 (prestin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene is a member of the SLC26A/SulP transporter family. It encodes a protein that is specifically expressed in outer hair cells (OHCs) of the cochlea and is essential in auditory processing. Intracellular anions are thought to act as extrinsic voltage sensors, which bind to this protein and trigger the conformational changes required for rapid length changes in OHCs. Mutations in this gene have been associated with non-syndromic hearing loss. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. (provided by RefSeq)

Immunogen

SLC26A5 (NP_945350, 645 a.a. ~ 741 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DFTQVNFIDSVGVKTLAGIVKEYGDVGIYVYLAGCSAQVVNDLTRNRFFENPALWELLFHSIHDAVLGSQLREALAEQEASAPPSQEDLEPNATPAT

Biochem/physiol Actions

Solute carrier family 26 member 5 (SLC26A5) functions as a transporter. Conformational changes in SLC26A5 leads to a change in the length of cochlear outer hair cells (OHC), which have a role in auditory perception.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seth L Alper et al.
Molecular aspects of medicine, 34(2-3), 494-515 (2013-03-20)
The phylogenetically ancient SLC26 gene family encodes multifunctional anion exchangers and anion channels transporting a broad range of substrates, including Cl(-), HCO3(-), sulfate, oxalate, I(-), and formate. SLC26 polypeptides are characterized by N-terminal cytoplasmic domains, 10-14 hydrophobic transmembrane spans, and
Jacob Pearson Keller et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(4), 1325-1332 (2014-01-24)
The solute carrier gene family 26 (SLC26) encodes membrane proteins with diverse physiological roles but with the common feature of halide involvement. Here, we present bioinformatic and biochemical evidence that SLC26 proteins have intrinsically disordered regions (IDRs) in their C-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service