Skip to Content
Merck
All Photos(4)

Key Documents

WH0009223M3

Sigma-Aldrich

Monoclonal Anti-MAGI1 antibody produced in mouse

clone 7B4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AIP3, Anti-BAIAP1, Anti-BAP1, Anti-MAGI1, Anti-TNRC19, Anti-WWP3, Anti-membrane associated guanylate kinase, WW and PDZ domain containing 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7B4, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGI1(9223)

Immunogen

MAGI1 (NP_004733, 761 a.a. ~ 859 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SHSTQVLPEFPPAEAQAPDQTDSSGQKKPDPFKIWAQSRSMYENRPMSPSPASGLSKGEREREINSTNFGECPIPDYQEQDIFLWRKETGFGFRILGGN

Application

Monoclonal Anti-MAGI1 antibody produced in mouse has been used for Western Blotting.

Biochem/physiol Actions

Membrane associated guanylate kinase, WW and PDZ domain containing 1 (MAGI1) acts as a scaffolding protein. It takes part in tight junction formation and binds to β-catenin to suppress the Wnt signaling pathway. The gene encoding it has been linked with schizophrenia and the protein is downregulated in hepatocellular carcinoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of interferon-? by MAGI-1 and its interaction with influenza A virus NS1 protein with ESEV PBM.
Kumar M, et.al
PLoS ONE, 7(7), e41251-e41251 (2012)
21942217
Derringer J
JAMA Psychiatry (Chicago, Ill.), 72(7), 642-650 (2015)
AmotL2 integrates polarity and junctional cues to modulate cell shape
Sara Hultin
Scientific Reports, 7 (2017)
The E-cadherin/AmotL2 complex organizes actin filaments required for epithelial hexagonal packing and blastocyst hatching
Sebastian H
Scientific Reports (2017)
Downregulation of MAGI1 associates with poor prognosis of hepatocellular carcinoma.
Zhang G, et.al
Journal of Investigative Surgery, 25(2), 93-99 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service