Skip to Content
Merck
All Photos(5)

Documents

WH0007337M1

Sigma-Aldrich

Monoclonal Anti-UBE3A antibody produced in mouse

clone 2F6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ANCR, Anti-AS, Anti-E6AP, Anti-EPVE6AP, Anti-HPVE6A, Anti-ubiquitin protein ligase E3A (human papilloma virus E6-associated protein, Angelman syndrome)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE3A(7337)

General description

This gene encodes an E3 ubiquitin-protein ligase, part of the ubiquitin protein degradation system. This imprinted gene is maternally expressed in brain and biallelically expressed in other tissues. Maternally inherited deletion of this gene causes Angelman Syndrome, characterized by severe motor and intellectual retardation, ataxia, hypotonia, epilepsy, absence of speech, and characteristic facies. The protein also interacts with the E6 protein of human papillomavirus types 16 and 18, resulting in ubiquitination and proteolysis of tumor protein p53. Alternative splicing of this gene results in three transcript variants encoding three isoforms with different N-termini. Additional transcript variants have been described, but their full length nature has not been determined. (provided by RefSeq)

Immunogen

UBE3A (AAH09271, 51 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ETFQQLITYKVISNEFNSRNLVNDDDAIVAASKCLKMVYYANVVGGEVDTNHNEEDDEEPIPESSELTLQELLGEERRNKKGPRVDPLETELGVKTLDCR

Biochem/physiol Actions

The ubiquitin protein ligase E3A (UBE3A) functions as E3 ligase in the ubiquitin-proteasome pathway and act as transcriptional coactivator of steroid hormone receptors such as progesterone, estrogen and other hormone receptors. During papilloma virus infection, ubiquitin protein ligase E3A (UBE3A), along with E6 viral protein, plays a vital role in degradation of the cell cycle regulatory protein p53. Anomalistic degradation of p53 can cause cervical cancer. Inactivation or deletion of UBE3A gene leads to the neurodevelopmental disorder Angelman syndrome (AS), while overexpression of maternal UBE3A gene is associated with autism pathogenesis. Therefore UBE3A level has to be properly controlled for normal brain development.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytically Important Residues of E6AP Ubiquitin Ligase Identified Using Acid-Cleavable Photo-Cross-Linkers.
Krist DT and Statsyuk AV
Biochemistry, 54(29), 4411-4414 (2015)
An Autism-Linked Mutation Disables Phosphorylation Control of UBE3A.
Yi JJ
Cell, 162(4), 795-807 (2015)
Epigenetic regulation of UBE3A and roles in human neurodevelopmental disorders.
LaSalle JM
Epigenomics, 7(7), 1213-1228 (2015)
The Angelman syndrome ubiquitin ligase localizes to the synapse and nucleus, and maternal deficiency results in abnormal dendritic spine morphology.
Dindot SV
Human Molecular Genetics, 17(1), 111-118 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service