Skip to Content
Merck
All Photos(5)

Key Documents

WH0007291M1

Sigma-Aldrich

Monoclonal Anti-TWIST1 antibody produced in mouse

clone 3E11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ACS3, Anti-BPES2, Anti-BPES3, Anti-SCS, Anti-TWIST, Anti-twist homolog 1 (acrocephalosyndactyly 3; Saethre-Chotzen syndrome) (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TWIST1(7291)

Related Categories

General description

Twist family bHLH transcription factor 1 (TWIST1) is a basic helix-loop-helix transcription factor and the gene encoding it is localized on human chromosome 7p21.1.

Immunogen

TWIST1 (NP_000465, 100 a.a. ~ 202 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PQSYEELQTQRVMANVRERQRTQSLNEAFAALRKIIPTLPSDKLSKIQTLKLAARYIDFLYQVLQSDELDSKMASCSYVAHERLSYAFSVWRMEGAWSMSASH

Biochem/physiol Actions

Twist family bHLH transcription factor 1 (TWIST1) has a role in the determination of cell lineage and differentiation during embryogenesis. During the development of neural crest, it is also involved in modulating cell movement and tissue reorganization. Mutations in the TWIST1 gene have been associated with Saethre-Chotzen syndrome (4) and the protein is also linked to various cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MACC1 ? more than metastasis?
Facts and predictions about a novel gene
Ulrike Stein
Journal of Molecular Medicine (2010)
Prognostic and clinicopathological value of Twist expression in breast cancer: A meta-analysis.
Qiao W
PLoS ONE (2017)
Saethre-Chotzen syndrome caused by TWIST 1 gene mutations: functional differentiation from Muenke coronal synostosis syndrome.
Kress W
European Journal of Human Genetics (2006)
Reactivation of TWIST1 contributes to Ewing sarcoma metastasis.
Pediatric Blood & Cancer (2018)
Ping Fan et al.
European journal of cancer (Oxford, England : 1990), 50(2), 457-468 (2013-11-05)
Our publications demonstrate that physiological concentrations of oestrogen (E2) induce endoplasmic reticulum and oxidative stress which finally result in apoptosis in E2-deprived breast cancer cells, MCF-7:5C. c-Src is involved in the process of E2-induced stress. To mimic the clinical administration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service