Skip to Content
Merck
All Photos(5)

Documents

WH0004790M1

Sigma-Aldrich

Monoclonal Anti-NFKB1 antibody produced in mouse

clone 2E6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp686C01211, Anti-EBP1, Anti-KBF1, Anti-MGC54151, Anti-NFKBp105, Anti-NFKBp50, Anti-NFkappaB, Anti-nuclear factor of kappa light polypeptide gene enhancer in B-cells 1 (p105)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKB1(4790)

General description

NFKB1 (nuclear factor κB subunit 1) is a dimeric transcription factor. It belongs to the REL family. This gene is located on human chromosome 4q24.

Immunogen

NFKB1 (AAH51765, 860 a.a. ~ 969 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDNYEVSGGTVRELVEALRQMGYTEAIEVIQAASSPVKTTSQAHSLPLSPASTRQQIDELRDSDSVCDSGVETSFRKLSFTESLTSGASLLTLNKMPHDYGQEGPLEGKI

Biochem/physiol Actions

NFKB1 (nuclear factor κB subunit 1) controls the maturation of NK (natural killer) cells and effector functions. This gene helps in the upregulation of intrauterine adhesion inflammatory factors, hence NFKB1 plays a major role in inflammatory diseases.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EBV induces persistent NF-?B activation and contributes to survival of EBV-positive neoplastic T- or NK-cells
Takada H, et al.
PLoS ONE, 12(3) (2017)
NFKB1 regulates human NK cell maturation and effector functions
Lougaris V, et al.
Clinical Immunology (Orlando, Fla.) (2017)
Elevated NF-?B signaling in Asherman syndrome patients and animal models
Wang X, et al.
Oncotarget, 8(9), 15399-15406 (2017)
J Spring
FEBS letters, 400(1), 2-8 (1997-01-02)
For the growing fraction of human genes with identified functions there are often homologues known from invertebrates such as Drosophila. A survey of well established gene families from aldolases to zinc finger transcription factors reveals that usually a single invertebrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service