Skip to Content
Merck
All Photos(4)

Key Documents

WH0001269M1

Sigma-Aldrich

Monoclonal Anti-CNR2 antibody produced in mouse

clone 3C7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CB2, Anti-CX5, Anti-cannabinoid receptor 2 (macrophage)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNR2(1269)

General description

Cannabinoid receptor 2 (CNR2) is encoded by the gene mapped to human chromosome 1p36.11. The encoded protein belongs to a novel cadherin-related multigene family. Members of this family contain six cadherin repeats and conserved DXNDNXPXF and DXD motifs with constitute putative calcium-binding sequences.
The cannabinoid delta-9-tetrahydrocannabinol is the principal psychoactive ingredient of marijuana. The proteins encoded by this gene and the cannabinoid receptor 1 (brain) (CNR1) gene have the characteristics of a guanine nucleotide-binding protein (G-protein)-coupled receptor for cannabinoids. They inhibit adenylate cyclase activity in a dose-dependent, stereoselective, and pertussis toxin-sensitive manner. These proteins have been found to be involved in the cannabinoid-induced CNS effects (including alterations in mood and cognition) experienced by users of marijuana. The cannabinoid receptors are members of family 1 of the G-protein-coupled receptors. (provided by RefSeq)

Immunogen

CNR2 (NP_001832.1, 302 a.a. ~ 360 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RSGEIRSSAHHCLAHWKKCVRGLGSEAKEEAPRSSVTETEADGKITPWPDSRDLDLSDC

Application

Monoclonal Anti-CNR2 antibody produced in mouse has been used in western blot analysis.

Biochem/physiol Actions

Cannabinoid receptor 2 (CNR2) plays a crucial role in mediating anti-inflammatory and anti-atherosclerotic effects in vivo. It is also involved in cancer, bone metabolism and pain perception. CNR2 might be implicated in selective cell–cell interaction. Mutations in the gene has been associated with trochanter and hip bone mineral density (BMD).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emerging therapeutic targets in cancer induced bone disease: A focus on the peripheral type 2 cannabinoid receptor.
Marino S and Idris A
Pharmacological Research, 119, 391-403 (2017)
Common polymorphisms in the cannabinoid CB2 receptor gene (CNR2) are not associated with myocardial infarction and cardiovascular risk factors.
Reinhard W, et al.
International Journal of Molecular Medicine, 22, 165-174 (2008)
Multiple osteoporosis susceptibility genes on chromosome 1p36 in Chinese.
Huang QY
Bone, 44, 984-988 (2009)
Role of the endocannabinoid system in the control of mouse myometrium contractility during the menstrual cycle
Pagano E, et al.
Biochemical Pharmacology, 124, 83-93 (2017)
Diversity Revealed by a Novel Family of Cadherins Expressed in Neurons at a Synaptic Complex
Kohmura N, et al.
Neuron, 20, 1137-1151 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service