Skip to Content
Merck
All Photos(1)

Key Documents

W3501

Sigma-Aldrich

Anti- Bovine Whey Proteins, antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Whey Proteins, bovine antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

bovine

technique(s)

immunoelectrophoresis: suitable
indirect ELISA: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Whey protein is obtained during cheese production. About 20% of the bovine milk consists of this natural protein.

Specificity

The product is reactive with bovine α-lactalbumin, sheep and goat whey proteins.

Application

Anti-Whey Proteins, bovine antibody produced in rabbit may be used in:
  • immunoelectrophoresis
  • immunodiffusion
  • enzyme-linked immunosorbent assay (ELISA)
  • PEPSCAN analysis

Biochem/physiol Actions

Whey protein might help in the reduction of long- and short-term appetite. It liberates various gut peptides, like, cholecystokinin and glucagon-like peptide 1. Whey protein supplementation is considered as a simple and feasible method to alleviate cancer cachexia syndrome. Whey protein subfractions possesses anti-cancer effects.
Whey proteins are globular proteins produced during cheese production. They are categorized as “fast” proteins as they are emptied faster from the stomach and enter the blood more rapidly than other proteins such as casein.

Physical form

Rabbit anti-Bovine Whey Proteins is provided as a liquid with 0.1% sodium azide added as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Williams et al.
Journal of immunological methods, 213(1), 1-17 (1998-07-22)
Two different epitope mapping techniques were used to identify linear epitopes recognised by polyclonal IgG antibodies from rabbits immunised with bovine beta lactoglobulin (BLG), which is generally regarded as a major allergen in milk. The first, PEPSCAN, was used to
Kevin D Tipton et al.
Medicine and science in sports and exercise, 36(12), 2073-2081 (2004-12-01)
Determination of the anabolic response to exercise and nutrition is important for individuals who may benefit from increased muscle mass. Intake of free amino acids after resistance exercise stimulates net muscle protein synthesis. The response of muscle protein balance to
Filipe J Teixeira et al.
Pharmacological research, 144, 245-256 (2019-04-22)
Cancer remains a public health challenge in the identification and development of ideal pharmacological therapies and dietary strategies. The use of whey protein as a dietary strategy is widespread in the field of oncology. The two types of whey protein
Mehdi Mollahosseini et al.
Clinical nutrition ESPEN, 20, 34-40 (2017-10-27)
Specific components of dairy, such as whey proteins may have beneficial effects on body composition by suppressing appetite, although the findings of existing studies have been inconsistent. Therefore, a meta-analysis of randomized controlled trials was performed to investigate effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service