Skip to Content
Merck
All Photos(1)

Key Documents

W1625

SAFC

Waymouth MB 752/1 Medium

With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Waymouth Medium, Waymouth′s medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Application

For the cultivation of mouse L929 cells in a serum-free environment. Also used in whole organ culture, establishment of carcinoma cell lines from pleural effusions, and the growth of potentially tumorigenic cells prior to their assessment in vitro.

Quantity

Formulated to contain 14.0 grams of powder per liter of medium.

Reconstitution

Supplement with 2.2 g/L sodium bicarbonate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Brendan D Stamper et al.
PloS one, 6(10), e26557-e26557 (2011-10-27)
Craniosynostosis is a disease defined by premature fusion of one or more cranial sutures. The mechanistic pathology of single-suture craniosynostosis is complex and while a number of genetic biomarkers and environmental predispositions have been identified, in many cases the causes
Gul Ozcan et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 21(6), 1552-1560 (2017-01-01)
Cancer is a long process that leads the organism to death and is associated with the normal cells acquiring the ability to divide permanently. Nowadays, the use of natural products in cancer therapy has a great importance. In addition, working
Behnam Abbasian et al.
mSphere, 4(5) (2019-09-06)
Urgency urinary incontinence (UUI) and overactive bladder (OAB) can both potentially be influenced by commensal and urinary tract infection-associated bacteria. The sensing of bladder filling involves interplay between various components of the nervous system, eventually resulting in contraction of the
Tala Shekarian et al.
Science translational medicine, 11(515) (2019-10-28)
Although immune checkpoint-targeted therapies are currently revolutionizing cancer care, only a minority of patients develop durable objective responses to anti-PD-1, PD-L1, and CTLA-4 therapy. Therefore, new therapeutic interventions are needed to increase the immunogenicity of tumors and overcome the resistance
Fu-Chi Kang et al.
Oncology reports, 41(6), 3565-3574 (2019-04-20)
In the western world, there is an increasing trend of occurrence in testicular cancer. Treatment of malignant testicular cancer is primarily combined surgery with various chemical drugs. Propofol has been frequently used as an anesthetic and sedative induction agent, which

Related Content

Waymouth MB 752/1 Medium Formulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service