Skip to Content
Merck
All Photos(1)

Key Documents

V9879

Sigma-Aldrich

[Arg8]-Vasopressin acetate salt

≥95% (HPLC)

Synonym(s):

β-Hypophamine, AVP, Antidiuretic hormone, Arginine vasopressin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H65N15O12S2 · xC2H4O2
CAS Number:
Molecular Weight:
1084.23 (free base basis)
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥95% (HPLC)

form

powder

solubility

water: 20.0-20.8 mg/mL, clear, colorless to faintly yellow

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(O)=O.N[C@H]1CSSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](Cc3ccc(O)cc3)NC1=O)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(N)=O

InChI

1S/C46H65N15O12S2.C2H4O2/c47-27-22-74-75-23-33(45(73)61-17-5-9-34(61)44(72)56-28(8-4-16-53-46(51)52)39(67)54-21-37(50)65)60-43(71)32(20-36(49)64)59-40(68)29(14-15-35(48)63)55-41(69)31(18-24-6-2-1-3-7-24)58-42(70)30(57-38(27)66)19-25-10-12-26(62)13-11-25;1-2(3)4/h1-3,6-7,10-13,27-34,62H,4-5,8-9,14-23,47H2,(H2,48,63)(H2,49,64)(H2,50,65)(H,54,67)(H,55,69)(H,56,72)(H,57,66)(H,58,70)(H,59,68)(H,60,71)(H4,51,52,53);1H3,(H,3,4)/t27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1

InChI key

ZHFGXNMADRRGQP-HPILINOVSA-N

Gene Information

Amino Acid Sequence

Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2 [Disulfide Bridge: 1-6]

General description

Arginine vasopressin is a nonapeptide.[1] It is produced by anterior hypothalamus nerve cells.[2]

Application

[Arg8]-Vasopressin acetate salt has been:
  • used as a vasopressin receptor agonist to test vasopressin receptors[3]
  • used to evoke peripheral V1a receptor-mediated vasoconstriction[4]
  • as a supplement in Ham′s F12/Dulbecco′s modified Eagle′s medium for proximal tubule and glomerular explant cell culture[5]

Biochem/physiol Actions

Endogenous antidiuretic hormone in most mammalian species. It is released into the blood from nerve terminals in the posterior pituitary and median eminence. Arg8-vasopressin is also a neurotransmitter in the central nervous system. It is implicated in a variety of physiological processes including diuresis, vasoregulation, and memory. Regulates water balance by antidiuretic action; contracts arterioles (vasopressor action).
Endogenous antidiuretic hormone in most mammalian species; neurotransmitter in the central nervous system implicated in a variety of physiological processes including diuresis, vasoregulation, and memory.

Preparation Note

[Arg8]-Vasopressin acetate salt dissolves water at 20 mg/ml to yield a clear colorless to faint yellow solution.

Other Notes

Lyophilized from 0.5% acetic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Voluntary locomotion linked with cerebral activation is mediated by vasopressin V1a receptors in free-moving mice
Masuki S, et al.
The Journal of Physiology, 591(14), 3651-3665 (2013)
Nucleotide sequence of cloned cDNA encoding bovine arginine vasopressin-neurophysin II precursor
Land H, et al.
Nature, 295(5847), 299-299 (1982)
Rare incorporation of bone marrow-derived cells into kidney after folic acid-induced injury
Szczypka MS, et al.
Stem Cells, 23(1), 44-54 (2005)
Medical Physiology, 424-424 (2009)
Daniel J van Wamelen et al.
Sleep, 36(1), 117-125 (2013-01-05)
To study whether sleep and circadian rhythm disturbances in patients with Huntington's disease (HD) arise from dysfunction of the body's master clock, the hypothalamic suprachiasmatic nucleus. Postmortem cohort study. Eight patients with HD and eight control subjects matched for sex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service