Skip to Content
Merck
All Photos(1)

Key Documents

V9134

Sigma-Aldrich

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse

clone KDR-1, ascites fluid

Synonym(s):

Anti-KDR, Anti-VEGF R-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

KDR-1, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry (frozen sections): 1:800 using human placenta
indirect ELISA: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)

General description

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (mouse IgG1 isotype) is derived from the KDR-1 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Vascular endothelial growth factor (VEGF), is a homodimeric heparin-binding glycoprotein. It is a member of a family of endothelial cell mitogenic and angiogenic factors. The VEGFR2 gene has been mapped to human chromosome 4q11-q12, which is the same locus for PDGF receptor and c-kit.
The antibody reacts specifically with VEGF receptor 2 (KDR) and does not recognize VEGF receptor-1 (Flt1 receptor), VEGF receptor 3 (sFlt4), and PDGF-Rβ.

Immunogen

recombinant human extracellular domain of VEGFR-2 (KDR).

Application

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse has been used in:
  • immunohistochemistry
  • histology
  • immunostaining
  • enzyme linked immunosorbent assay (ELISA)

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse was used for FACS analysis of HUVECs.

Biochem/physiol Actions

Vascular endothelial growth factor (VEGF) specifically stimulates the proliferation of endothelial cells isolated from both small and large vessels. These include endothelial cells from adrenal cortex, cerebral cortex, fetal and adult aorta and human umbilical vein.
Vascular endothelial growth factors receptors (VEGF Rs) are receptor tyrosine kinases that bind to VEGF. VEGF plays an important role in embryonic vasculogenesis, angiogenesis and homeostasis. The binding of VEGF to VEGF R2 results in activation of MAPK, PI3K, PKC, FAK and Src kinase pathways. The biological roles of VEGF R2 include in epithelial cell growth, proliferation, survival, migration, vascular permeability and angiogenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chie-Pein Chen et al.
Stem cells (Dayton, Ohio), 26(2), 550-561 (2007-11-03)
Maternal cells can become engrafted in various fetal organs during pregnancy. The nature of the cells and the mechanisms of maternofetal cell trafficking are not clear. We demonstrate that human lineage-negative, CD34-negative (Lin(-)CD34(-)) multipotent mesenchymal stromal cells express alpha(2), alpha(4)
Axitinib affects cell viability and migration of a primary foetal lung adenocarcinoma culture
Menna C, et al.
Cancer Investigation, 32(1), 13-21 (2014)
Vascular endothelial growth factor (VEGF) is an autocrine growth factor for VEGF receptor-positive human tumors
Masood R, et al.
Blood, 98(6), 1904-1913 (2001)
Neuropilin-1 promotes VEGFR-2 trafficking through Rab11 vesicles thereby specifying signal output
Ballmer-Hofer K, et al.
Blood, 118(3), 816-826 (2011)
Lidia Sinka et al.
Blood, 119(16), 3712-3723 (2012-01-28)
Adult-type lympho-myeloid hematopoietic progenitors are first generated in the aorta-gonad-mesonephros region between days 27 and 40 of human embryonic development, but an elusive blood forming potential is present earlier in the underlying splanchnopleura. In the present study, we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service