Skip to Content
Merck
All Photos(1)

Key Documents

UC167

Sigma-Aldrich

Oxidized Nifedipine

powder, ~95% (HPLC)

Synonym(s):

2,6-Dimethyl-4-(2´-nitrophenyl)-3,5-pyridinecarboxylic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O6
CAS Number:
Molecular Weight:
344.32
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

~95% (HPLC)

form

powder

color

yellow

mp

100-105 °C

storage temp.

2-8°C

SMILES string

COC(=O)c1c(C)nc(C)c(C(=O)OC)c1-c2ccccc2[N+]([O-])=O

InChI

1S/C17H16N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8H,1-4H3

InChI key

UMQHJQGNGLQJPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxidized Nifedipine may be used:
  • to study enzyme kinetics using UV high-performance liquid chromatography (UV-HPLC)
  • as a standard in calibration curve preparation in high-performance liquid chromatography (HPLC) for the quantification of cytochrome P450 3A4 (CYP3A4)-mediated nifedipine oxidation in human microsomes
  • in nifedipine metabolism activity assay of Hep G2 cells transfected with hepatocyte nuclear factor-1α(HNF1α)

Oxidized nifedipine has been used for apoptotic signaling studies in dopaminergic neurons.

Biochem/physiol Actions

CYP3A4 nifedipine metabolite. Nifedipine (parent compound) is an antianginal and antihypertensive agent.
Nifedipine is a dihydropyridine derivative and a calcium channel blocker. It is used in treating hypertension and angina pectoris. Oxidized Nifedipine is an oxidation product of nifedipine generated either by exposure to UV or day light. The enzyme cytochrome P450 3A (CYP3A4) metabolizes nifedipine.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel A de Erausquin et al.
Molecular pharmacology, 63(4), 784-790 (2003-03-20)
We describe a new molecular mechanism of cell death by excitotoxicity mediated through nuclear transcription factor kappa B (NF kappa B) in rat embryonic cultures of dopaminergic neurons. Treatment of mesencephalic cultures with alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) resulted in a number
CYP3As catalyze nifedipine oxidation in pig liver microsomes: Enzyme kinetics, inhibition and functional expression
Jiang J, et al.
Catalysis Communications, 12(8), 694-697 (2011)
Wiriyaporn Sumsakul et al.
Asian Pacific journal of tropical medicine, 8(11), 914-918 (2015-11-29)
To investigate the propensity of plumbagin to inhibit the three isoforms of human cytochrome P450 (CYP), i.e., CYP1A2, CYP2C19, and CYP3A4 using human liver microsomes in vitro. Inhibitory effects of plumbagin on the three human CYP isoforms were investigated using pooled
D G Waller et al.
British journal of clinical pharmacology, 18(6), 951-954 (1984-12-01)
Oral administration of nifedipine (20 and 30 mg tablets) to six volunteers was associated with a bioavailability of 0.43 and the presence of its nitropyridine analogue in the plasma. This metabolite was present in only trace amounts in samples taken
J L Born et al.
Chemical research in toxicology, 2(1), 57-59 (1989-01-01)
The primary deuterium isotope effect on Vm for the microsomal oxidation of the dihydropyridine calcium entry blocker nifedipine [4-(2-nitrophenyl)-2,6-dimethyl-3,5- bis(methoxycarbonyl)-1,4-dihydropyridine] has been measured. The magnitude of the kinetic isotope effect, 6.7, suggests that the rate-limiting step in the mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service