Skip to Content
Merck
All Photos(2)

Key Documents

U5377

Sigma-Aldrich

Uracil 1-β-D-arabinofuranoside

lymphoma antiproliferative

Synonym(s):

Arauridine, Spongouridin, 1-β-D-Arabinofuranosyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
Beilstein:
28749
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7+,8-/m1/s1

InChI key

DRTQHJPVMGBUCF-CCXZUQQUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-β-d-arabinofuranosyluracil (Ara-U) is the detoxification product or metabolite of 1-β-d-arabinofuranosylcytosine (Ara-C). Uracil 1-β-D-arabinofuranoside is the human metabolite of a cytostatic agent called cytarabine (CYT).

Application

Uracil 1-β-D-arabinofuranoside may be used as a reference compound in the analysis of cytostatics and metabolites in hospital wastewater by hydrophilic interaction chromatography/tandem mass spectrometry to demonstrate its analytical possibilities and limitations. It may also be used as a standard for chemical analysis to investigate the ecotoxicity and genotoxicity of cytotoxic antineoplastic drugs, like cytarabine (CYT) and their metabolites, like uracil-1-β-d-arabinofuranoside (AraU).

Biochem/physiol Actions

1-β-D-Arabinofuranosyluracil (ara-U) is capable of inhibiting proliferation of L5178Y mouse lymphoma cells at 17 μM concentration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metabolism of 1-beta-D-arabinofuranosyluracil in mouse L5178Y cells.
W E Müller et al.
Cancer research, 39(3), 1102-1107 (1979-03-01)
Radka Zounkova et al.
Chemosphere, 81(2), 253-260 (2010-07-14)
In spite of growing scientific concern about pharmaceuticals in the environment, there is still a lack of information especially with regard to their metabolites. The present study investigated ecotoxicity and genotoxicity of three widely used cytostatic agents 5-fluorouracil (5-FU), cytarabine
Melissa N McCracken et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(5), 1857-1862 (2013-01-16)
Positron emission tomography (PET) reporter genes allow noninvasive whole-body imaging of transplanted cells by detection with radiolabeled probes. We used a human deoxycytidine kinase containing three amino acid substitutions within the active site (hdCK3mut) as a reporter gene in combination
Hwi Young Kim et al.
PloS one, 7(11), e50377-e50377 (2012-12-05)
Modeling of short-term viral dynamics of hepatitis B with traditional biphasic model might be insufficient to explain long-term viral dynamics. The aim was to develop a novel method of mathematical modeling to shed light on the dissociation between early and
Kai Chen et al.
Nuclear medicine and biology, 39(7), 1019-1025 (2012-04-17)
[(18)F]-FMAU is a PET tracer being evaluated for imaging cell proliferation. Current multi-step procedures of [(18)F]-FMAU synthesis are time-consuming, resulting in low radiochemical yield and inconvenient applications for the clinic. We have previously reported the use of Friedel-Crafts catalysts for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service