Skip to Content
Merck

TRUE3

TrueGel3D Hydrogel Kits

FAST-DEXTRAN, allow cell recovery, CD cell-degradable crosslinker

Synonym(s):

Gel kits

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Storage temp.:
−70°C
Shipped in:
dry ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

storage temp.

−70°C

General description

The gel is formed by stable thioether bonds between maleimide groups of dextran (FAST-DEXTRAN) and thiol groups on CD cell-degradable crosslinker. Malemide-functionalized dextran is specifically used to decrease the time for gel formation. The dextran polymer does not have cell attachment sites, but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1UM) prior to the crosslinking step. The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala), which allows cells to spread and migrate by secreting matrix metalloproteases (MMP1, MMP3, MMP7 and MMP9).

TrueGel3D enzymatic cell recovery solution (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR, western blots and other molecular biological studies.
TrueGel3D® provides a high degree of control in a 3D hydrogel environment, providing greater consistency. It enables to fine-tune the microenvironments around the tissues or cells under study.

Application

TRUE3-1KT is used for cell migration studies of amoeboid cells including neutrophils, dendritic cells and lymphocytes. It has been used to study the effect of mechanic stimulus over tumor development, tumor progression and cell differentiation.

Features and Benefits

  • Fast gelling (takes few seconds to minutes after addition of crosslinker)
  • Contains MMP-cleavable crosslinker
  • Allow cell recovery and post-culture analyses
  • Flexibility in modulating gel stiffness
  • Cells do not attach to hydrogel (if not customized with TrueGel3D RGD peptide)

Other Notes

TRUE3-1KT contains lyophilized FAST-DEXTRAN (polymer) and CD cell-degradable crosslinker with reconstitution buffer, TrueGel3D thioglycerol and water.

Legal Information

TRUEGEL3D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Avoid prolonged exposure of CD cell-degradable crosslinker to air.


Still not finding the right product?

Explore all of our products under TrueGel3D Hydrogel Kits


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library