Skip to Content
Merck
All Photos(1)

Key Documents

TOX2

Sigma-Aldrich

In Vitro Toxicology Assay Kit, XTT based

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

form

solid

usage

 kit sufficient for 1,000 tests

packaging

pkg of 1 kit

storage condition

dry at room temperature

λmax

450 nm

application(s)

cell analysis
detection

detection method

colorimetric

storage temp.

−20°C

Application

For spectrophotometric measurement of cell viability by mitochodrial dehydrogenase. Absorbance of converted dye is measured at a wavelength of 450 nm.

Biochem/physiol Actions

Conversion of XTT to a water-soluble colored formazan derivative.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond W Wong et al.
PLoS pathogens, 9(3), e1003241-e1003241 (2013-04-05)
To develop new approaches to control HIV-1 replication, we examined the capacity of recently described small molecular modulators of RNA splicing for their effects on viral RNA metabolism. Of the drugs tested, digoxin was found to induce a dramatic inhibition
Catherine Gerard et al.
Cell transplantation, 21(1), 127-137 (2011-05-04)
Duchenne muscular dystrophy (DMD) is the most frequent muscular dystrophy in children and young adults. Currently, there is no cure for the disease. The transplantation of healthy myoblasts is an experimental therapeutic strategy, since it could restore the expression of
O S Weislow et al.
Journal of the National Cancer Institute, 81(8), 577-586 (1989-04-19)
We have developed an effective and optimally safe microculture method for rapid and convenient assay of the in vitro cytopathic effects of human immunodeficiency virus (HIV-1) on human lymphoblastoid or other suitable host cells. The assay procedure is applicable to
Queenie P Vong et al.
Blood, 124(26), 3905-3913 (2014-10-30)
Thymocyte selection-associated high mobility group box protein family member 2 (TOX2) is a transcription factor belonging to the TOX family that shares a highly conserved high mobility group DNA-binding domain with the other TOX members. Although TOX1 has been shown
Eunsohl Lee et al.
Journal of cellular biochemistry, 117(12), 2815-2824 (2016-05-07)
Prostate cancer (PCa) is known to develop resistance to chemotherapy. Growth arrest-specific 6 (GAS6), plays a role in tumor progression by regulating growth in many cancers. Here, we explored how GAS6 regulates the cell cycle and apoptosis of PCa cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service