Skip to Content
Merck
All Photos(2)

Key Documents

T9262

Sigma-Aldrich

Tamoxifen citrate salt

≥99% (TLC), powder, PKC inhibitor

Synonym(s):

(Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29NO · C6H8O7
CAS Number:
Molecular Weight:
563.64
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

product name

Tamoxifen citrate salt, ≥99%

Assay

≥99%

solubility

methanol: soluble 50 mg/mL, clear, colorless

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(c1ccccc1)=C(/c2ccccc2)c3ccc(OCCN(C)C)cc3

InChI

1S/C26H29NO.C6H8O7/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,4,19-20H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;

InChI key

FQZYTYWMLGAPFJ-OQKDUQJOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tamoxifen has been used for analyzing its effects on radiation sensitivity and steroid receptor content in human breast cancer cells. It has also been used for studying the accumulation of triacylglycerol in hepatic tissues of female rats. Furthermore, studies have reported that environmental magnetic fields can block antiproliferative functions of tamoxifen in human breast cancer cells.

Biochem/physiol Actions

Protein kinase C inhibitor. Induces apoptosis in human malignant glioma cell lines. Tamoxifen and its metabolite 4-hydroxytamoxifen are selective estrogen response modifiers (SERMs) that act as estrogen antagonists in mammary gland. Blocks estradiol-stimulated VEGF production in breast tumor cells.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Tamoxifen citrate salt dissolves in methanol at 50 mg/ml to yield a clear, colorless solution.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G H Paulsen et al.
Acta oncologica (Stockholm, Sweden), 35(8), 1011-1019 (1996-01-01)
Possible influences of tamoxifen and estradiol on in vitro radiation sensitivity and cellular receptor content after irradiation and/or tamoxifen treatment were studied in breast cancer cell lines; estrogen receptor (ER) and progesterone receptor (PgR) positive cell lines MCF-7 and MCF-7/TAM(R)-1
J D Harland et al.
Bioelectromagnetics, 18(8), 555-562 (1997-01-01)
We have previously reported that environmental-level magnetic fields (1.2 microT [12 milligauss], 60 Hz) block the growth inhibition of the hormone melatonin (10(-9) M) on MCF-7 human breast cancer cells in vitro. We now report that the same 1.2 microT
Oddrun Anita Gudbrandsen et al.
Journal of lipid research, 47(10), 2223-2232 (2006-07-26)
Tamoxifen can induce hepatic steatosis in women. In this study, we wanted to elucidate the mechanism behind the tamoxifen-induced accumulation of triacylglycerol in liver in female rats, and we hoped to prevent this development by combination treatment with the modified
Kevin S Hughes et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(19), 2382-2387 (2013-05-22)
To determine whether there is a benefit to adjuvant radiation therapy after breast-conserving surgery and tamoxifen in women age ≥ 70 years with early-stage breast cancer. Between July 1994 and February 1999, 636 women (age ≥ 70 years) who had
P Fox et al.
British journal of cancer, 109(1), 147-153 (2013-06-20)
The host inflammatory response has a vital role in carcinogenesis and tumour progression. We examined the prognostic value of inflammatory markers (albumin, white-cell count and its components, and platelets) in pre-treated patients with advanced renal cell carcinoma (RCC). Using data

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service