Skip to Content
Merck
All Photos(1)

Key Documents

T8532

Sigma-Aldrich

Triton X-100

suitable for electrophoresis

Synonym(s):

t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

form

liquid

mol wt

micellar avg mol wt 80,000
average mol wt 625

aggregation number

100-155

technique(s)

DNA extraction: suitable
DNA purification: suitable
electrophoresis: suitable

color

APHA: ≤100

turbidity

≤5.0 NTU

CMC

0.2-0.9 mM (20-25°C)

transition temp

cloud point 65 °C
pour point ~7 °C

solubility

water: soluble

density

1.06 g/mL at 25 °C (lit.)

HLB

13.5

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Detergent used for isoelectric focusing (IEF) and two-dimensional electrophoresis. Concentrations between 1-4% (v/v) are typically used in an IEF gel. Also used for cell permeabilization and protein solubilization including high molecular mass proteins.

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

also commonly purchased with this product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allison E Songstad et al.
Stem cells translational medicine, 6(6), 1533-1546 (2017-05-06)
Age-related macular degeneration (AMD) is a leading cause of irreversible blindness in the Western world. Although, the majority of stem cell research to date has focused on production of retinal pigment epithelial (RPE) and photoreceptor cells for the purpose of
Leonard Herrfurth et al.
Frontiers in molecular neuroscience, 10, 2-2 (2017-02-15)
Vascular endothelial growth factor (VEGF) is well known as the growth factor with wide-ranging functions even in the central nervous system (CNS). Presently, most attention is given to the investigation of its role in neuronal protection, growth and maturation processes
Friederike Pfeiffer et al.
Acta neuropathologica, 122(5), 601-614 (2011-10-11)
In experimental autoimmune encephalomyelitis (EAE), an animal model for multiple sclerosis (MS), loss of the blood-brain barrier (BBB) tight junction (TJ) protein claudin-3 correlates with immune cell infiltration into the CNS and BBB leakiness. Here we show that sealing BBB
Jing Cui et al.
Experimental eye research, 88(3), 438-444 (2008-11-27)
Previous investigators reported that epiretinal membranes isolated from patients with proliferative vitreoretinopathy (PVR) express various platelet-derived growth factor (PDGF) family members and PDGF receptors (PDGFRs) (Cui, J.Z., Chiu, A., Maberley, D., Ma, P., Samad, A., Matsubara, J.A., 2007. Stage specificity
Adam A Strömstedt et al.
Antimicrobial agents and chemotherapy, 53(2), 593-602 (2008-11-26)
Methods for increasing the proteolytic stability of EFK17 (EFKRIVQRIKDFLRNLV), a new peptide sequence with antimicrobial properties derived from LL-37, were evaluated. EFK17 was modified by four d-enantiomer or tryptophan (W) substitutions at known protease cleavage sites as well as by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service