Skip to Content
Merck
All Photos(5)

Key Documents

T7505

Sigma-Aldrich

Tetramethylammonium hydroxide pentahydrate

≥97%

Synonym(s):

TMAH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(OH) · 5H2O
CAS Number:
Molecular Weight:
181.23
Beilstein:
3714235
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

vapor density

1.1 (vs air)

description

non-ionic

Assay

≥97%

autoignition temp.

878 °F

mol wt

181.23 g/mol

expl. lim.

36 %

mp

67-70 °C (lit.)

SMILES string

O.O.O.O.O.[OH-].C[N+](C)(C)C

InChI

1S/C4H12N.6H2O/c1-5(2,3)4;;;;;;/h1-4H3;6*1H2/q+1;;;;;;/p-1

InChI key

MYXKPFMQWULLOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylammonium hydroxide (TMAOH) is an ion-pairing agent that is usually used in analytical chemistry. It is used in the analysis of marine compounds by thin-layer chromatography-pyrolysis-gas chromatography-mass spectrometry (TLC-pyrolysis-GC/MS) and thermal desorption/capillary GC/inductively coupled plasma mass spectrometry. TMAOH is used in the capillary zone electrophoresis technique for the separation of inorganic anions. Some studies report the oligonucleotide reassociation in the presence of TMAOH. TMAOH aids in the synthesis of organic and inorganic compounds, such as vinyl halides and tetramethylammonium dithiocarboxylates. It is useful in the preparation of iron oxide nanoparticles for use as magnetic resonance imaging contrast agents.

Application

Tetramethylammonium hydroxide pentahydrate has been used in a study to assess strains of Pseudomnas species by a single-step gas chromatographic characterization procedure. It has also been used in a study to investigate solid-state metal hydride batteries nickel oxyhydroxide-metal hydride and manganese dioxide-metal hydride.
Tetramethylammonium hydroxide pentahydrate has been used in the synthesis of silicalite-1 seed crystals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1 Dermal - STOT SE 1

Target Organs

Central nervous system, Liver,thymus

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Wade et al.
Applied microbiology, 27(2), 303-311 (1974-02-01)
Strains of saccharolytic and nonsaccharolytic Pseudomonas species were examined by a new single-step gas chromatographic characterization procedure. Cells were digested in a methanolic solution of tetramethylammonium hydroxide pentahydrate, and the digestates were subjected to gas-liquid chromatographic analysis. The chromatograms were
ZuLiang Chen et al.
Analytical and bioanalytical chemistry, 375(1), 182-187 (2003-01-10)
In this paper, the use of an ion-pairing reagent to improve the separation selectivity of inorganic anions in CZE was demonstrated by the addition of tetramethylammonium hydroxide (TMAOH) to the electrolyte. The separation of inorganic anions (Cl(-), I(-), Br(-), NO(2)(-)
High conductivity and phase transitions of tetramethylammoniumhydroxidepentahydrate, (CH3)4NOH?5H2O
Kuriyama, N., et al.
Solid State Ionics, 40-41, 906-906 (1990)
Solid-state metal hydride batteries using tetramethylammoniumhydroxidepentahydrate
Kuriyama, N., et al.
Solid State Ionics, 53-56, 688-688 (1992)
J Vercauteren et al.
Analytical chemistry, 73(7), 1509-1514 (2001-04-26)
The extraction and preconcentration capabilities of a new extraction technique, stir bar sorptive extraction, were combined with the separation power of capillary gas chromatography (CGC) and the low limits of detection (LODs) of inductively coupled plasma mass spectrometry (ICPMS) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service