Skip to Content

T6830

Thyroglobulin human

Synonym(s):

Thyroglobulin human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-721-3
NACRES:
NA.56
MDL number:
Form:
lyophilized solid
Assay:
≥95% (SDS-PAGE)
Biological source:
human
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

≥95% (SDS-PAGE)

form

lyophilized solid

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... TG(7038)

General description

Human Thyroglobulin (Tg) is a 660 kDa, dimeric protein produced by and used entirely within the thyroid gland. Human Thyroglobin is used by the thyroid gland to produce the thyroid hormones thyroxine (T4) and triiodothyryonine. Patients with Hashimoto′s thyroiditis or Graves′ disease, frequently develop antibodies against human Thyroglobulin. Thryoglobulin levels in the blood can be used as a tumor marker for certain kinds of thyroid cancer and in Graves′ disease.

Biochem/physiol Actions

Thyroglobulin has been shown to induce expression of platelet activating factor in human peripheral blood mononuclear cells which is found elevated in some patients with thyroid disease.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library