T5905
L-Thyronine
≥98.0% (TLC)
Synonym(s):
3-(p-[p-Hydroxyphenoxy]phenyl)-L-alanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H15NO4
CAS Number:
Molecular Weight:
273.28
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
L-Thyronine,
Assay
≥98.0% (TLC)
Quality Level
form
powder
SMILES string
N[C@@H](Cc1ccc(Oc2ccc(O)cc2)cc1)C(O)=O
InChI
1S/C15H15NO4/c16-14(15(18)19)9-10-1-5-12(6-2-10)20-13-7-3-11(17)4-8-13/h1-8,14,17H,9,16H2,(H,18,19)/t14-/m0/s1
InChI key
KKCIOUWDFWQUBT-AWEZNQCLSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D B Ramsden et al.
Biomedical mass spectrometry, 11(4), 193-198 (1984-04-01)
The mass spectral properties of four classes of derivatives of thyronine are discussed ( oxazolidinone , O-methyl oxazolidinone , O-acetyl oxazolidinone and N,O- diheptafluorobutyryl methyl ester). An assay for thyronine in human urine is described based on the N,O- diheptafluorobutyryl
I J Chopra et al.
The Journal of clinical endocrinology and metabolism, 75(1), 189-194 (1992-07-01)
A highly sensitive, specific, and reproducible RIA has been developed to measure T3 sulfate (T3S). Only T4 sulfate cross-reacted significantly (approximately 3%) in the RIA; rT3 sulfate, T4, T3, rT3, and diiodothyronine cross-reacted less than 0.01%. T3S was bound by
J Köhrle
Cellular and molecular life sciences : CMLS, 57(13-14), 1853-1863 (2001-02-24)
Thyroid hormones control growth, development, differentiation and metabolism in vertebrates. Most of the actions of the active thyroid hormone T3 (3,5,3'-triiodo-L-thyronine) are exerted via ligand-activated nuclear T3 receptors. Activation of the secretory product of the thyroid gland, L-thyroxine (3,3',5,5'-tetraiodo-L-thyronine), or
J S LoPresti et al.
The Journal of clinical endocrinology and metabolism, 70(5), 1479-1484 (1990-05-01)
A hidden pool of rT3 production represents a source of rT3 that is minimally reflected in circulating rT3 levels. To test for the existence of such a source of rT3 production in man, varying doses of the generalized deiodinase inhibitor
Martin A Paquette et al.
PloS one, 9(6), e101155-e101155 (2014-06-28)
Thyroid hormone (TH) exerts its effects by binding to the thyroid hormone receptor (TR), which binds to TH response elements (TREs) to regulate target gene expression. We investigated the relative ability of liganded homodimers TR and retinoid X receptor (RXR)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service