Skip to Content
Merck
All Photos(6)

Documents

T5625

Sigma-Aldrich

(±)-α-Lipoic acid

synthetic, ≥99% (titration), powder

Synonym(s):

(±)-1,2-Dithiolane-3-pentanoic acid, 6,8-Dithiooctanoic acid, DL-α-Lipoic acid, DL-6,8-Thioctic acid, Lip(S2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2S2
CAS Number:
Molecular Weight:
206.33
Beilstein:
81853
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

description

Oxidized form

Assay

≥99% (titration)

form

powder

solubility

ethanol: soluble 50 mg/mL

SMILES string

OC(=O)CCCCC1CCSS1

InChI

1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)

InChI key

AGBQKNBQESQNJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-LA can reinstate endothelial activities and alleviate oxidative stress in high-fat fed diabetic rats4. It has also been used to analyze ROS production in astrocytes5.

Biochem/physiol Actions

Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.

Preparation Note

(±)-α-Lipoic acid (α-LA) is soluble in ethanol at 50 mg/ml

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniely V S Costa et al.
Pharmaceuticals (Basel, Switzerland), 13(11) (2020-11-07)
Irinotecan, an anticancer drug, induces diarrhea and intestinal inflammation, resulting in an increase in the cost of care and in treatment delays. In this study, we investigated whether alpha-lipoic acid (α-LA) could improve irinotecan-mediated intestinal inflammation, diarrhea and dysmotility. Intestinal
Massimo Dal Monte et al.
Diagnostics (Basel, Switzerland), 10(3) (2020-03-07)
Melatoninergic agents are known to reduce intraocular pressure (IOP). The present study was performed to evaluate the effect of nanomicellar formulations of melatoninergic agents on IOP in the rat. Tonometry was used to measure IOP in eyes instilled with melatonin
Fatma M Ghoneim et al.
Biology, 9(9) (2020-08-23)
Sodium valproate (VPA) is an antiepileptic drug (AED) licensed for epilepsy and used during pregnancy in various indications. Alpha-lipoic acid (ALA) is a natural compound inducing endogenous antioxidant production. Our study aimed to investigate the effect of prenatal administration of
Reda Hassan Elbakary et al.
Journal of microscopy and ultrastructure, 8(2), 42-50 (2020-08-09)
Polychlorinated biphenyl (PCB) is considered one of the environmental pollutants. It is used as hydraulic coils in vacuum pumps, pesticides transformers, heat-exchange systems, capacitors and as additives in adhesive inks, paints, plastics, copying paper and sealants. Alpha lipoic acid (ALA)
Maria C N Marchetto et al.
Cell stem cell, 3(6), 649-657 (2008-12-02)
Amyotrophic lateral sclerosis (ALS) is a neurodegenerative disease characterized by motor neuron death. ALS can be induced by mutations in the superoxide dismutase 1 gene (SOD1). Evidence for the non-cell-autonomous nature of ALS emerged from the observation that wild-type glial

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service