Skip to Content
Merck
All Photos(1)

Key Documents

T3573

Sigma-Aldrich

RANK Ligand/TRANCE human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Osteoclast Differentiation Factor (ODF), Osteoprotegerin Ligand (OPGL), Receptor Activator of NF-KB Ligand (RANKL), TNF-related activation-induced cytokines (TRANCE)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

1.5-7.5 ng/mL ED50

mol wt

glycolysylated recombinant product ~35 kDa by SDS-PAGE
calculated mol wt 23 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

Gene Information

human ... TNFSF11(8600)

General description

Tumor necrosis factor (ligand) superfamily, member 11 (Tnfsf11) is also known as receptor activator of nuclear factor-κB ligand (RANKL). It is a type II transmembrane signaling receptor. It has a molecular weight of around 35kDa. This protein contains an amino-terminal intracellular tail and a carboxy-terminal extracellular region, that bears a connecting stalk and a receptor-binding domain. Tnfsf11 is located on human chromosome 13q14.11.

Application

RANK Ligand/TRANCE human can be used in in vitro osteoclastogenic assays for osteoclast differentiation.
RANK Ligand/TRANCE human has been used to stimulate differentiation.

Biochem/physiol Actions

Member of the TNF superfamily; induces activation of the c-jun N-terminal kinase, enhances T-cell growth and dendritic cell function, induces osteoclastogenesis, and lymph node organogenesis; cell surface receptor for RANK Ligand.
RANK Ligand (receptor activator of NF-kB ligand (RANKL) also referred to as TNF-related activation induced cytokines (TRANCE) or osteoprotegerin ligand is a protein that belongs to tumor necrosis factor (TNF) family. It stimulates the mature dendritic cells and induces the cytokine production. RANKL also facilitates bone remodelling and possess an angiogenic activity.
RANK Ligand/TRANCE, a member of the TNF superfamily, induces activation of the c-jun N-terminal kinase, enhances T-cell growth and dendritic cell function, induces osteoclastogenesis, and lymph node organogenesis. Mouse and human RANK Ligand share 85% amino acid identity. RANK is the cell surface receptor for RANK Ligand.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20 mM MOPS and 500 mM NaCl, pH 6.5, with 50 μg BSA per 1 μg as a carrier protein.

Analysis Note

The biological activity is measured by its ability to induce osteoclast differentiation on mouse splenocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Josien et al.
Journal of immunology (Baltimore, Md. : 1950), 162(5), 2562-2568 (1999-03-11)
TNF-related activation-induced cytokine (TRANCE) is a member of the TNF family recently identified in activated T cells. We report here that TRANCE mRNA is constitutively expressed in memory, but not naive, T cells and in single-positive thymocytes. Upon TCR/CD3 stimulation
Cytosolic phospholipase A2 and eicosanoids modulate life, death and function of human osteoclasts in vitro
Allard-Chamard H, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 90(4), 117-123 (2014)
RANKL employs distinct binding modes to engage RANK and the osteoprotegerin decoy receptor
Nelson C A, et al.
Structure, 20(11), 1971-1982 (2012)
Jeong-Ki Min et al.
The Journal of biological chemistry, 278(41), 39548-39557 (2003-08-02)
Vascular endothelial growth factor (VEGF) is known as a key regulator of angiogenesis during endochondral bone formation. Recently, we demonstrated that TNF-related activation-induced cytokine (TRANCE or RANKL), which is essential for bone remodeling, also had an angiogenic activity. Here we
Matrine prevents bone loss in ovariectomized mice by inhibiting RANKL-induced osteoclastogenesis
Chen X, et al.
Faseb Journal, 31(11), 4855-4865 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service